Merck
  • Pharmacokinetic study of hydroxypropylmethylcellulose microparticles loaded with cimetidine.
Pharmacokinetic study of hydroxypropylmethylcellulose microparticles loaded with cimetidine.
Advances in clinical and experimental medicine : official organ Wroclaw Medical University (2013-03-08)
Rozina Kousar, Mahmood Ahmad, Ghulam Murtaza, Shujaat A Khan, Sabiha Karim, Izhar Hussain
ABSTRACT
The objective of this study was to assess the pharmacokinetic behavior of floating hydroxypropylmethylcellulose microparticles loaded with cimetidine (FMC) prepared using the non-solvent addition coacervation technique. Based on the physico-chemical characteristics of three formulations (FMC1, FMC2 and FMC3), FMC2 having a 1:3 ratio of cimetidine:HPMC was found optimum. For in vivo analysis, a new HPLC analytical method was developed and validated. The optimized formulations were subjected to in vivo studies to calculate the various pharmacokinetic parameters for developed optimized microparticulate formulation FMC3. The developed floating microparticles of cimetidine were further evaluated by in vivo experimentation. The bioavailability parameters were found as: Cmax 1508.79 ± 37.95 ng/ml, Tmax 3.67 ± 0.17 h and AUC 14366.19 ± 377.64 ng h /mL. For prolonged drug release in the stomach, developed floating microparticles of cimetidine (FMC3) may be used, thereby improving the bioavailability and patient compliance.
Product Description
Supelco
Cimetidine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methyl cellulose, viscosity 3000-5500 mPa.s, 2 % in H2O(20 °C)
Sigma-Aldrich
Methyl cellulose, viscosity: 15 cP
Sigma-Aldrich
Methyl cellulose, 27.5-31.5% (Methoxyl content), viscosity: 400 cP
Sigma-Aldrich
Methyl cellulose, viscosity: 1,500 cP
Sigma-Aldrich
Methyl cellulose, viscosity: 4,000 cP
Sigma-Aldrich
Methyl cellulose, viscosity: 25 cP
Sigma-Aldrich
Methyl cellulose, 26.0-33.0% (Methoxy group (dry basis)), meets USP testing specifications, viscosity: 1,500 cP
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, viscosity 2,600-5,600 cP, 2 % in H2O(20 °C)(lit.)
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, viscosity 40-60 cP, 2 % in H2O(20 °C)(lit.)
Sigma-Aldrich
Hypromellose, meets USP testing specifications
Sigma-Aldrich
Methyl cellulose, 26.0-33.0% (methoxyl group, on Dry Basis), meets USP testing specifications, viscosity: 400 cP
Sigma-Aldrich
Methyl cellulose, 27.5-31.5% methoxyl basis
Sigma-Aldrich
Methyl cellulose, tested according to Ph. Eur.
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, average Mn ~10,000
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, average Mn ~90,000
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, average Mn ~86,000
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, average Mn ~120,000
Sigma-Aldrich
Methyl cellulose, medium viscosity, Methoxyl content 27.5-31.5 %
Sigma-Aldrich
Cimetidine
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, viscosity 80-120 cP, 2 % in H2O(20 °C)(lit.)
Sigma-Aldrich
Methyl cellulose, viscosity: 15 cP, BioReagent, suitable for cell culture