Merck
  • Preparation of primary amine derivatives of the magic-size nanocluster (CdSe)13.
Preparation of primary amine derivatives of the magic-size nanocluster (CdSe)13.
Inorganic chemistry (2013-03-06)
Yuanyuan Wang, Yi-Hsin Liu, Ying Zhang, Paul J Kowalski, Henry W Rohrs, William E Buhro
ABSTRACT
Four [(CdSe)13(RNH2)13] derivatives (R = n-propyl, n-pentyl, n-octyl, and oleyl) are prepared by reaction of Cd(OAc)2·2H2O and selenourea in the corresponding primary-amine solvent. Nanoclusters grow in spontaneously formed amine-bilayer templates and are characterized by elemental analysis, IR spectroscopy, UV-vis spectroscopy, TEM, and low-angle XRD. Derivative [(CdSe)13(n-propylamine)13] is isolated as a yellowish-white solid (MP 98 °C) on the gram scale. These compounds are the first derivatives of magic-size CdSe nanoclusters to be isolated in purity.
Sigma-Aldrich
Cadmium selenide, -325 Mesh particle size, 99.99% trace metals basis, electronic grade
Pricing and availability is not currently available.