Merck
  • Heavy metal and phenol adsorptive properties of biochars from pyrolyzed switchgrass and woody biomass in correlation with surface properties.
Heavy metal and phenol adsorptive properties of biochars from pyrolyzed switchgrass and woody biomass in correlation with surface properties.
Journal of environmental management (2013-03-05)
Yanxue Han, Akwasi A Boateng, Phoebe X Qi, Isabel M Lima, Jianmin Chang
ABSTRACT
In this work, the surface structures of biochars, derived from three types of biomass, switchgrass (SG), hardwood (HW) and softwood (SW) through either fast pyrolysis (FP) in a fluidized-bed reactor (at 500 °C) or slow pyrolysis (at 500° and 700 °C), were studied in detail, and compared with that of the activated carbons obtained by steam activation of the slow pyrolyzed biochars (at 500 °C). The surface acidic functional groups were determined quantitatively by the Boehm Titration method. The adsorptive properties of heavy metals, Zn(2+) and Cu(2+) onto the biochars and the activated carbons were investigated by the adsorption isotherms and SEM images, and correlated with the surface properties. ATR-FTIR and GC techniques were used to analyze the adsorptive behavior of phenol onto the biochars and activated carbons, and the results demonstrated that phenol adsorption capability is directly proportional to the micropore surface area as well as the combined level of the accessible carboxylic and lactonic groups. The relative adsorption capacity with respect to the biomass precursor follows the order: SW > HW > SG.
Product Description
Supelco
Phenol solution, 5000 μg/mL in methanol, certified reference material
Supelco
Phenol solution, certified reference material, 500 μg/mL in methanol
Sigma-Aldrich
Phenol
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Phenol solution, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Sigma-Aldrich
Phenol solution, BioReagent, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Supelco
Phenol, PESTANAL®, analytical standard
Sigma-Aldrich
Phenol, puriss., ≥99.5% (GC), meets analytical specification of Ph. Eur., BP, USP, crystalline (detached)
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Supelco
Phenol solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
Sigma-Aldrich
Phenol, BioUltra, TE-saturated, ~73% (T)
Sigma-Aldrich
Phenol, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Phenol, ACS reagent, ≥99.0%
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Supelco
Phenol, Pharmaceutical Secondary Standard; Certified Reference Material