Merck
  • Larval settlement and metamorphosis of the mussel Mytilus coruscus in response to monospecific bacterial biofilms.
Larval settlement and metamorphosis of the mussel Mytilus coruscus in response to monospecific bacterial biofilms.
Biofouling (2013-03-05)
Jin-Long Yang, Pei-Jing Shen, Xiao Liang, Yi-Feng Li, Wei-Yang Bao, Jia-Le Li
ABSTRACT
The effects of bacterial biofilms (BFs) on larval settlement and metamorphosis of the mussel, Mytilus coruscus, were investigated in the laboratory. Of nine different isolates, Shewanella sp.1 BF induced the highest percentage of larval settlement and metamorphosis, whereas seven other isolates had a moderate inducing activity and one isolate, Pseudoalteromonas sp. 4, had a no inducing activity. The inducing activity of individual bacterial isolates was not correlated either with their phylogenetic relationship or with the surfaces from which they were isolated. Among the eight bacterial species that demonstrated inducing activity, bacterial density was significantly correlated with the inducing activity for each strain, with the exception of Vibrio sp. 1. The Shewanella sp. 1 BF cue that was responsible for inducing larval settlement and metamorphosis was further investigated. Treatment of the BFs with formalin, antibiotics, ultraviolet irradiation, heat, and ethanol resulted in a significant decrease in their inducing activities and cell survival. BF-conditioned water (CW) did not induce larval metamorphosis, but it triggered larval settlement behavior. A synergistic effect of CW with formalin-fixed Shewanella sp. 1 BF significantly promoted larval metamorphosis. Thus, a cocktail of chemical cues derived from bacteria may be necessary to stimulate larval settlement and metamorphosis in this species.
Sigma-Aldrich
Penicillin G sodium salt, ~1650 U/mg
Sigma-Aldrich
Penicillin G sodium salt, 96.0-102.0%
Sigma-Aldrich
Penicillin G sodium salt, powder, BioReagent, suitable for cell culture
Supelco
Penicillin G potassium salt, VETRANAL®, analytical standard
Sigma-Aldrich
Penicillin G potassium salt, 95.0 - 102.0%
Sigma-Aldrich
Penicillin G potassium salt
Sigma-Aldrich
Penicillin G potassium salt, powder, BioReagent, suitable for cell culture