Merck
Degradation of malachite green by dielectric barrier discharge plasma.
Water science and technology : a journal of the International Association on Water Pollution Research (2013-02-19)
P Manoj Kumar Reddy, B Ramaraju, Ch Subrahmanyam
ABSTRACT
Oxidative decomposition of aqueous organic pollutant malachite green (MG) was studied in a dielectric barrier discharge reactor operated under ambient conditions. Total organic carbon content analysis confirmed the mineralization of the pollutant leading to the formation of carbon dioxide, which was confirmed by an infrared analyzer. Typical results indicated that the degradation rate increases with increasing applied voltage and decreases with increasing concentration. Dye degradation followed first order kinetics. The intermediate products formed during the degradation of MG were identified by a high resolution mass spectrometer (HR-MS) and proposed a plausible mechanism for the mineralization process.
Sigma-Aldrich
Malachite Green solution, for microscopy
Sigma-Aldrich
Malachite Green oxalate salt, for microscopy, crystalline, S. No.: 754
Sigma-Aldrich
Malachite Green oxalate salt, certified by the Biological Stain Commission
Supelco
Malachite Green oxalate salt, VETRANAL®, analytical standard
Sigma-Aldrich
Malachite Green oxalate salt, Technical grade
Sigma-Aldrich
Schaeffer and Fulton Spore Stain Solution A, for microscopy