Merck
  • Rapid determination of sumatriptan in human plasma by ultra performance liquid chromatography-tandem mass spectrometry and its application to clinical pharmacokinetic study.
Rapid determination of sumatriptan in human plasma by ultra performance liquid chromatography-tandem mass spectrometry and its application to clinical pharmacokinetic study.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2013-02-16)
Jeong Ju Seo, Jeonghyeon Park, Min Ho Bae, Mi-sun Lim, Sook Jin Seong, Joomi Lee, Sung Min Park, Hae Won Lee, Young-Ran Yoon
ABSTRACT
A sensitive and simple detection method coupling ultra-performance liquid chromatography with tandem mass spectrometry was developed and validated to analyze sumatriptan levels in human plasma. The plasma sample preparations for the analysis were based on liquid-liquid extraction with ethyl acetate, evaporation, and reconstitution. MS/MS detection was performed on a triple-quadrupole tandem mass spectrometer by monitoring the protonated parent→daughter ion pairs at m/z 296→58 and m/z 388→71 for sumatriptan and terazosin (internal standard), respectively. The method was validated with respect to its specificity, linearity, sensitivity, accuracy, precision, recovery, and stability. The calibration curve was linear from 0.5 to 50 ng/mL (r>0.999). The mean extraction recovery for sumatriptan was higher than 62.3%. The method accuracy was within 97.4%, and the relative standard deviation of the intra- and inter-day precision values was within 11.7% at all quality control levels. Plasma samples that contained sumatriptan were stable under three freeze-thaw cycles, short- and long-term storage, and autosampler conditions. This method was successfully applied to a pharmacokinetic study conducted with 10 healthy volunteers. After oral administration of 50-mg sumatriptan and serial blood sampling over 12 h, the mean area under the plasma concentration-time curve from time 0 to 12 h and the maximum plasma concentration were 116.2 ng h/mL and 33.2 ng/mL, respectively.
Sigma-Aldrich
Sumatriptan succinate, ≥98% (HPLC), solid
Sumatriptan succinate, European Pharmacopoeia (EP) Reference Standard
Sumatriptan for system suitability, European Pharmacopoeia (EP) Reference Standard