Merck
  • Straw N-halamines: evaluation in single and multistage filtration systems.
Straw N-halamines: evaluation in single and multistage filtration systems.
Carbohydrate polymers (2013-02-13)
Abd El-Shafey I Ahmed, Gabriel Cavalli, Michael E Bushell, John N Wardell, Steve Pedley, Katarina Charles, John N Hay
ABSTRACT
New N-halamines (I-Cl and II-Cl) based on cellulose extracted from rice straw have been evaluated in single and multistage filtration systems against bacteria and viruses. Escherichia coli and Staphylococcus aureus were used as examples of Gram-negative and Gram-positive bacteria respectively while PRD1 bacteriophage was used as an example for viruses. II-Cl has achieved 9 log reductions in viable counts against E. coli in 2 h and S. aureus in 1h while it has achieved 7 log reductions against PRD1 in 5 h. The particle size of prepared materials was modified as well as the flow rate through the filtration systems. The antimicrobial activity of modified cellulose was proved to be comparable to some synthetic biocidal polymers from the same type in similar water treatment systems.
Sigma-Aldrich
Calcium chloride dihydrate, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Calcium chloride dihydrate, ACS reagent, ≥99%
Sigma-Aldrich
Calcium chloride dihydrate, puriss., meets analytical specification of Ph. Eur., USP, FCC, E509, 99-103%, ≤0.0001% Al
Sigma-Aldrich
Calcium chloride dihydrate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Calcium chloride dihydrate, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Calcium chloride dihydrate, BioXtra, ≥99.0%
Sigma-Aldrich
Calcium chloride dihydrate, for molecular biology, ≥99.0%
Sigma-Aldrich
Alginic acid sodium salt from brown algae, Medium viscosity
Sigma-Aldrich
Alginic acid sodium salt from brown algae, BioReagent, suitable for immobilization of micro-organisms
Sigma-Aldrich
Calcium chloride dihydrate, tested according to Ph. Eur.
Sigma-Aldrich
Calcium chloride dihydrate, BioUltra, for molecular biology, ≥99.5% (KT)
Supelco
Calcium standard for AAS, analytical standard, 1.000 g/L Ca+2 in hydrochloric acid, traceable to BAM
Sigma-Aldrich
Alginic acid calcium salt from brown algae
Sigma-Aldrich
Alginic acid sodium salt, powder
Sigma-Aldrich
Alginic acid sodium salt from brown algae, low viscosity
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Alginic acid from brown algae, powder
Sigma-Aldrich
Calcium chloride
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Sodium alginate
Sigma-Aldrich
Calcium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Supelco
Calcium ion solution for ISE, 0.1 M Ca, analytical standard (for ion-selective electrodes)