Merck
Natural porous agar materials from macroalgae.
Carbohydrate polymers (2013-02-13)
Matteo Francavilla, Antonio Pineda, Carol S K Lin, Massimo Franchi, Pasquale Trotta, Antonio A Romero, Rafael Luque
ABSTRACT
Porous agar materials have been prepared from marine macroalgae species using a simple microwave-assisted extraction/drying methodology, providing a new family of polysaccharide derived porous solids. The microwave-assisted extraction allows a more efficient and less time-consuming extraction of the polysaccharide compared to conventional extraction protocols based on conventional heating. DRIFT and (13)C NMR results indicated that the internal agar structure (based on d-galactose and 3,6-anhydro-l-galactose linked units) was preserved after the extraction methodology, which opens a wide range of future possibilities and applications for this new family of porous polysaccharides. The extracted agar materials, which have already applications per se due to their high purities, could be subsequently transformed into a novel family of attractive mesoporous agar materials that could be used as natural templates for the production of nanocrystals of metal oxides.
Sigma-Aldrich
Agar, ash 2.0-4.5%
Sigma-Aldrich
Agar, suitable for plant cell culture, powder
Sigma-Aldrich
Agar, Type A, suitable for plant cell culture
Millipore
Agar, powder, suitable for microbiology
Millipore
Agar, suitable for microbiology
Sigma-Aldrich
Agar, suitable for plant cell culture
Sigma-Aldrich
Agar, Type M, suitable for plant cell culture
Sigma-Aldrich
Agar, High gel strength, suitable for plant cell culture
Sigma-Aldrich
Select agar
Sigma-Aldrich
Agar, microbiology tested, suitable for plant cell culture, suitable for cell culture, powder
Sigma-Aldrich
Noble agar
Sigma-Aldrich
Agar, Bacteriological, microbiology tested, suitable for plant cell culture, granular
Millipore
Agar, suitable for microbiology
Millipore
Agar, high purity, low ionic content, low gel strenght, suitable for microbiology
Sigma-Aldrich
Bacteriological agar, for molecular biology