Merck
  • Three-dimensional homochiral manganese-lanthanide frameworks based on chiral camphorates with multi-coordination modes.
Three-dimensional homochiral manganese-lanthanide frameworks based on chiral camphorates with multi-coordination modes.
Chemical communications (Cambridge, England) (2013-02-12)
Dong-Bin Dang, Bing An, Yan Bai, Guang-Shui Zheng, Jing-Yang Niu
ABSTRACT
A series of 3D homochiral manganese-lanthanide frameworks have been synthesized based on chiral camphoric acid. In the heterometallic features, D-camphoric acids are unprecedentedly embedded in three coordination modes.
Product Description
Supelco
D-Camphor, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
(1S)-(−)-Camphor, 95%
Sigma-Aldrich
D-Camphor, ≥97%, FG
Camphor (dl), primary reference standard
Sigma-Aldrich
(±)-Camphor, meets analytical specification of Ph. Eur., BP, ≥95% (GC), racemic
Supelco
(−)-Camphor, analytical standard
Sigma-Aldrich
Camphor, 96%
Sigma-Aldrich
(±)-Camphor, purum, synthetic, ≥95.0% (GC)
Sigma-Aldrich
(±)-Camphor, ≥95.5%
Sigma-Aldrich
(1R)-(+)-Camphor, 98%
Camphor (racemic), European Pharmacopoeia (EP) Reference Standard