Merck
  • Effect of surface treatment on bond strength of Ti-10Ta-10Nb to low-fusing porcelain.
Effect of surface treatment on bond strength of Ti-10Ta-10Nb to low-fusing porcelain.
The Journal of prosthetic dentistry (2013-02-12)
Bo-Ah Lee, Ok-Su Kim, Mong-Sook Vang, Yeong-Joon Park
ABSTRACT
Ti-10Ta-10Nb alloy is a promising alloy for metal ceramic crowns because of its good corrosion resistance and low cytotoxicity. However, more information is needed on the bond strength between this alloy and porcelain. The purpose of this study was to compare the surface morphology, surface roughness, and bond strength of a Ti-10Ta-10Nb alloy, pure Ti, and a Ti-6Al-4V alloy. Ti-10Ta-10Nb, pure Ti, and Ti-6Al-4V specimens (25 × 3 × 0.55 mm plate) were prepared and then divided into 6 groups (n=8) according to surface treatment. Group P (control group) was polished with SiC paper. Groups S50 and S250 were airborne-particle abraded with 50 μm and 250 μm aluminum oxide powder. Group HCl was immersed in 10% HCl aqueous solution, and Group HF was immersed in 17% HNO(3)/HF solution. Group TiN was coated with TiN. Atomic force microscopy was used to observe the surface roughness of the metal surface. Scanning electron microscopy was used to analyze the surface profile. A 3-point bending test was performed to evaluate the bond strength. Two-way analysis of variance (ANOVA) was performed to compare the roughness and bond strength and statistical differences were revealed by the Bonferroni post hoc test (α=.05). There were significant differences in the surface roughness, surface profile, and bond strength of the Ti alloys according to the surface treatments. The groups with the higher mean surface roughness showed higher bond strength, but surface profile had a larger effect on the bond strength than surface roughness. Moreover, the bond strength of the Ti-10Ta-10Nb alloy was high. Ti-10Ta-10Nb would be more suitable for a metal ceramic crown than pure Ti or Ti-6Al-4V, which have limited use because of their low bond strength to porcelain.
Sigma-Aldrich
Tantalum, foil, thickness 0.05 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Tantalum, powder, −325 mesh, 99.9% trace metals basis
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Supelco
Hydrogen chloride – ethanol solution, ~1.25 M HCl, for GC derivatization, LiChropur
Sigma-Aldrich
Silicon carbide, nanopowder, <100 nm particle size
Sigma-Aldrich
Silicon carbide, -200 mesh particle size
Sigma-Aldrich
Silicon carbide, −400 mesh particle size, ≥97.5%
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Titanium nitride, <3 μm
Sigma-Aldrich
Hydrochloric acid, puriss., 24.5-26.0%
Sigma-Aldrich
Hydrofluoric acid, ACS reagent, 48%
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Hydrochloric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., fuming, ≥37%, APHA: ≤10
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Hydrofluoric acid, 48 wt. % in H2O, ≥99.99% trace metals basis
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Nitric acid, ACS reagent, ≥90.0%
Supelco
Nitric acid concentrate, 0.1 M HNO3 in water (0.1N), eluent concentrate for IC
Sigma-Aldrich
Nitric acid, 70%, purified by redistillation, ≥99.999% trace metals basis
Sigma-Aldrich
Tantalum, foil, thickness 0.5 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Tantalum, wire, diam. 1.0 mm, ≥99.9% trace metals basis