Merck
  • Conformational altered p53 affects neuronal function: relevance for the response to toxic insult and growth-associated protein 43 expression.
Conformational altered p53 affects neuronal function: relevance for the response to toxic insult and growth-associated protein 43 expression.
Cell death & disease (2013-02-09)
L Buizza, C Prandelli, S A Bonini, A Delbarba, G Cenini, C Lanni, E Buoso, M Racchi, S Govoni, M Memo, D Uberti
ABSTRACT
The role of p53 in neurodegenerative diseases is essentially associated with neuronal death. Recently an alternative point of view is emerging, as altered p53 conformation and impaired protein function have been found in fibroblasts and blood cells derived from Alzheimer's disease patients. Here, using stable transfected SH-SY5Y cells overexpressing APP751wt (SY5Y-APP) we demonstrated that the expression of an unfolded p53 conformation compromised neuronal functionality. In particular, these cells showed (i) augmented expression of amyloid precursor protein (APP) and its metabolites, including the C-terminal fragments C99 and C83 and β-amyloid peptide (ii) high levels of oxidative markers, such as 4-hydroxy-2-nonenal Michael-adducts and 3-nitro-tyrosine and (iii) altered p53 conformation, mainly due to nitration of its tyrosine residues. The consequences of high-unfolded p53 expression resulted in loss of p53 pro-apoptotic activity, and reduction of growth-associated protein 43 (GAP-43) mRNA and protein levels. The role of unfolded p53 in cell death resistance and lack of GAP-43 transcription was demonstrated by ZnCl(2) treatment. Zinc supplementation reverted p53 wild-type tertiary structure, increased cells sensitivity to acute cytotoxic injury and GAP-43 levels in SY5Y-APP clone.
Sigma-Aldrich
Zinc chloride solution, 1.9 M in 2-methyltetrahydrofuran
Sigma-Aldrich
3-Nitro-L-tyrosine, crystalline
Sigma-Aldrich
Zinc chloride, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Zinc chloride, AnhydroBeads, amorphous, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Zinc chloride, AnhydroBeads, amorphous, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Zinc chloride, anhydrous, powder, ≥99.995% trace metals basis
Sigma-Aldrich
Zinc chloride, 99.999% trace metals basis
Sigma-Aldrich
Zinc chloride 0.1 M solution
Sigma-Aldrich
Anti-APP A4 Antibody, a.a. 66-81 of APP {NT}, clone 22C11, clone 22C11, Chemicon®, from mouse
Sigma-Aldrich
Zinc chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Zinc chloride, puriss., meets analytical specification of Ph. Eur., BP, USP, 98-100.5%
Sigma-Aldrich
Zinc chloride, ACS reagent, ≥97%
Sigma-Aldrich
Zinc chloride, reagent grade, ≥98%
Sigma-Aldrich
Zinc chloride, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Zinc chloride solution, 0.5 M in THF