Merck
  • Androgens concentrations and second-to fourth-digit ratio (2D:4D) in girls with congenital adrenal hyperplasia (21-hydroxylase deficiency).
Androgens concentrations and second-to fourth-digit ratio (2D:4D) in girls with congenital adrenal hyperplasia (21-hydroxylase deficiency).
Neuro endocrinology letters (2013-02-09)
Joanna M Oswiecimska, Agnieszka Ksiazek, Katarzyna Sygulla, Magdalena Pys-Spychala, Grzegorz R Roczniak, Wojciech Roczniak, Malgorzata Stojewska, Katarzyna Ziora
ABSTRACT
Excessive hyperandrogenism, though proper hydrocortisone supplementation is a frequent clinical problem in girls with congenital adrenal hyperplasia (CAH). This may result from autonomic regulation of androgen production established in prenatal life. It has been suggested that the length of the second finger relative to the length of the fourth finger (2D;4D ratio) is negatively related to prenatal testosterone concentration. The retrospective study aimed to establish the relationship between the level of androgenization in utero determined using 2D:4D ratio and serum androgen concentrations in treated girls with CAH (21-OH deficiency) has been performed on 19 girls with CAH (21-OH deficiency) at the age of 3.7-19 years (mean 13.8 ± 4.07 years). All subjects were adequately treated with hydrocortisone (10-19 mg/m2; mean 13.81 ± 4.07 mg/m2). Anthropometric measurements of digits length were performed in all girls on X-rays obtained for bone age estimation. Apart from it, serum androgens concentrations (testosterone, androstenedione, s-DHEA) and 17-OH-progesterone (17-OHP) were assayed. Mean androgens serum concentrations in examined group were: testosterone 150.21 ± 155.44 ng/ml; androstenedione 4.15 ± 5.32 ng/ml, s-DHEA 70.39 ± 85.52 µg/dl. Mean 2D:4D ratio was 0.96 ± 0.04. Analysis of correlation showed positive linear correlations between testosterone, s-DHEA and 2D:4D ratio (r=0.53, p=0.023 and r=0.53; p=0.019, respectively). 2D:4D ratio parameter may be a simple test in indentification of female CAH patients prone to excessive androgen secretion despite proper treatment. The autonomization of adrenal androgens production in foetal life may cause its elevated levels in female patients with CAH although treated adequately.
Product Description
Supelco
4-Androstene-3,17-dione, VETRANAL®, analytical standard
Sigma-Aldrich
17α-Hydroxyprogesterone, ≥95%