Merck
  • Interventional embolization of giant thoracic tumors before surgical resection.
Interventional embolization of giant thoracic tumors before surgical resection.
Acta radiologica (Stockholm, Sweden : 1987) (2013-02-05)
Feng-yong Liu, Mao-qiang Wang, Qing-sheng Fan, Feng Duan, Zhi-jun Wang, Peng Song
ABSTRACT
Preoperative embolization of tumors is a well-established procedure that has been successfully applied in various clinical situations. Preoperative embolization can reduce the vascularity of tumors resulting in a clearer operative field, less difficult dissection, decreased blood loss, and, in some cases, a decrease in tumor size. However, few studies have been conducted regarding the preoperative embolization of giant thoracic tumors. To examine the effectiveness and safety of interventional embolization of giant thoracic tumors before surgical resection. A total of 14 consecutive patients with giant thoracic tumors received angiography and the feeding arteries of the tumors were embolized using polyvinyl alcohol (PVA) particles and gelatin sponges 1 day before surgical resection. The patient records were retrospectively reviewed and data regarding diagnoses, embolization, and surgical resection were recorded. Angiography revealed the feeding arteries of the tumors to be characterized by multiple branches and thickened vessel trunks with abnormal distal branches superimposed of the tumor shadow. Embolization was successfully without complications in all patients, and all feeding vessels of each tumor were occluded. Embolization reduced the severity of bleeding during surgery and decreased the difficulty of resection of the tumor. No intraoperative or postoperative complications occurred. Interventional embolization is a safe and efficient method to facilitate the surgical resection of giant thoracic tumors.
Sigma-Aldrich
Kuraray Poval ®56-98, Mw ~195,000
Sigma-Aldrich
Mowiol® 40-88, average Mw ~205,000 g/mol
Sigma-Aldrich
Mowiol® 18-88, Mw ~130,000
Sigma-Aldrich
Mowiol® 28-99, Mw ~145,000
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 146,000-186,000, 87-89% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), Mw 31,000-50,000, 98-99% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), Mw 13,000-23,000, 87-89% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), Mw 8,000-10,000, 80% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), Mw 85,000-124,000, 99+% hydrolyzed
Sigma-Aldrich
Mowiol® 10-98, Mw ~61,000
Sigma-Aldrich
Mowiol® 6-98, Mw ~47,000
Sigma-Aldrich
Mowiol® 4-88, Mw ~31,000
Sigma-Aldrich
Poly(vinyl alcohol), Mw 146,000-186,000, 99+% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 85,000-124,000, 87-89% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 13,000-23,000, 98% hydrolyzed
Sigma-Aldrich
Mowiol® 4-98, Mw ~27,000
Sigma-Aldrich
Mowiol® 8-88, Mw ~67,000
Sigma-Aldrich
Poly(vinyl alcohol), Fully hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), 87-90% hydrolyzed, average mol wt 30,000-70,000
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 31,000-50,000, 87-89% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), Mw 89,000-98,000, 99+% hydrolyzed
Sigma-Aldrich
Mowiol® 20-98, Mw ~125,000
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 130,000, 99+% hydrolyzed