Merck
  • Modifying excitation light dose of novel photosensitizer PVP-Hypericin for photodynamic diagnosis and therapy.
Modifying excitation light dose of novel photosensitizer PVP-Hypericin for photodynamic diagnosis and therapy.
Journal of photochemistry and photobiology. B, Biology (2013-02-05)
Rozhin Penjweini, Hans G Loew, Maria Eisenbauer, Karl W Kratky
ABSTRACT
Conventional photodynamic diagnosis (PDD) and therapy (PDT) makes use of photosensitizers that are excited by continuous light irradiation of specific wavelengths. In the case of PDT, the overdose of continuous excitation may lead to an expansion of necrosis in cancer cells or morbidity in healthy surroundings. The present study involves 5-h fluorescence imaging of living human lung epithelial carcinoma cells (A549) in the presence of a novel photosensitizer, PVP-Hypericin (PVP: polyvinylpyrrolidone) to optimize the excitation light doses for PDD and PDT. A number of time-lapse imaging experiments were performed using a low-power blue LED operating in either continuous or pulsed mode. The irradiances I(*) were 1.59, 6.34 and 14.27mW/cm(2), the pulse lengths L being 0.127, 1.29, 13, 54.5, 131 and 60,000ms. Then, the relation between irradiance, various exposure times, photobleaching and phototoxicity of PVP-Hyperycin was investigated. Results showed a nonlinear relationship between the amounts of excitation dose, cell viability and toxicity. For all experimental I(*), minimal phototoxicity and photobleaching was detected when cells were exposed to brief pulses of light (L⩽13ms). On the other hand, pulsed excitation with I(*)=14.27mW/cm(2) and L=131ms induced high percentages of apoptosis comparable to the long exposures of L=60,000ms and the continuous excitation. Thus, replacement of continuous excitation by a pulsed method seems applicable for PDT.
Sigma-Aldrich
Polyvinylpyrrolidone solution, K 60, 45% in H2O
Sigma-Aldrich
Polyvinylpyrrolidone, K 90
Sigma-Aldrich
Polyvinylpyrrolidone, K 30
Sigma-Aldrich
Polyvinylpyrrolidone, powder, average Mw ~29,000
Sigma-Aldrich
Polyvinylpyrrolidone, powder, average Mw ~55,000
Sigma-Aldrich
Polyvinylpyrrolidone, average Mw ~1,300,000 by LS
Sigma-Aldrich
Perylene, sublimed grade, ≥99.5%
Sigma-Aldrich
Perylene, ≥99%
Sigma-Aldrich
Polyvinylpyrrolidone, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Polyvinylpyrrolidone, suitable for plant cell culture, average mol wt 10,000
Sigma-Aldrich
Polyvinylpyrrolidone, mol wt (number average molecular weight Mn 360kDa)
Sigma-Aldrich
Polyvinylpyrrolidone, for molecular biology, nucleic acid hybridization tested, mol wt 360,000
Sigma-Aldrich
Polyvinylpyrrolidone, average mol wt 40,000
Sigma-Aldrich
Polyvinylpyrrolidone, average mol wt 10,000
Sigma-Aldrich
Polyvinylpyrrolidone, K 25, tested according to Ph. Eur.
Sigma-Aldrich
Hypericin from Hypericum perforatum, ~95% (HPLC)
Hypericin, primary reference standard
Sigma-Aldrich
Kollidon® 25
Sigma-Aldrich
Povidone, meets USP testing specifications
Povidone, European Pharmacopoeia (EP) Reference Standard
Supelco
Povidone, Pharmaceutical Secondary Standard; Certified Reference Material