Merck
  • Analysis of naturally-occurring radionuclides in coal combustion fly ash, gypsum, and scrubber residue samples.
Analysis of naturally-occurring radionuclides in coal combustion fly ash, gypsum, and scrubber residue samples.
Health physics (2013-01-31)
Angela R Roper, Michael G Stabin, Rossane C Delapp, David S Kosson
ABSTRACT
Coal combustion residues from coal-fired power plants can be advantageous for use in building and construction materials. These by-products contain trace quantities of naturally occurring radionuclides from the uranium and thorium series, as well as other naturally occurring radionuclides such as K. Analysis was performed on samples of coal fly ash, flue gas desulfurization, gypsum and scrubber sludges, fixated scrubber sludges, and waste water filter cakes sampled from multiple coal-fired power plants in the United States. The radioactive content of U and Th decay series nuclides was determined using gamma photopeaks from progeny Pb at 352 keV and Tl at 583 keV, respectively; K specific activities were determined using the 1,461 keV photopeak. The samples were hermetically sealed to allow for secular equilibrium between the radium parents and the radon and subsequent progeny. Samples were analyzed in a common geometry using two high purity germanium photon detectors with low energy detection capabilities. The specific activities (Bq kg) were compared to results from literature studies including different building materials and fly ash specific activities. Fly ash from bituminous and subbituminous coals had U specific activities varying from 30-217 Bq kg (mean + 1 s.d. 119 ± 45 Bq kg) and 72-209 Bq kg (115 ± 40 Bq kg), respectively; Th specific activities from 10-120 Bq kg (73 ± 26 Bq kg) and 53-110 Bq kg (81 ± 18 Bq kg), respectively; and K specific activities from 177 to 928 Bq kg (569 ± 184 Bq kg) and 87-303 Bq kg (171 ± 69 Bq kg), respectively. Gypsum samples had U, Th, and K specific activities approximately one order of magnitude less than measured for fly ash samples.
Sigma-Aldrich
Calcium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Calcium sulfate dihydrate, puriss., meets analytical specification of NF, E 516, 99.0-101.0% (based on anhydrous substance)
Sigma-Aldrich
Calcium sulfate dihydrate, ACS reagent, 98%
Sigma-Aldrich
Calcium sulfate, −325 mesh, 99%
Sigma-Aldrich
Calcium sulfate dihydrate, ReagentPlus®, ≥99%
Sigma-Aldrich
Drierite, with indicator, 10-20 mesh
Sigma-Aldrich
Calcium sulfate hemihydrate, purum, ≥97.0%
Sigma-Aldrich
Drierite, with indicator, 4 mesh
Sigma-Aldrich
Drierite, with indicator, 8 mesh
Sigma-Aldrich
Drierite, without indicator, 8 mesh
Sigma-Aldrich
Drierite, without indicator, 4 mesh
Sigma-Aldrich
Drierite, with indicator, 6 mesh