Merck
  • Glytube: a conical tube and parafilm M-based method as a simplified device to artificially blood-feed the dengue vector mosquito, Aedes aegypti.
Glytube: a conical tube and parafilm M-based method as a simplified device to artificially blood-feed the dengue vector mosquito, Aedes aegypti.
PloS one (2013-01-24)
André Luis Costa-da-Silva, Flávia Rosa Navarrete, Felipe Scassi Salvador, Maria Karina-Costa, Rafaella Sayuri Ioshino, Diego Soares Azevedo, Desirée Rafaela Rocha, Camila Malta Romano, Margareth Lara Capurro
ABSTRACT
Aedes aegypti, the main vector of dengue virus, requires a blood meal to produce eggs. Although live animals are still the main blood source for laboratory colonies, many artificial feeders are available. These feeders are also the best method for experimental oral infection of Ae. aegypti with Dengue viruses. However, most of them are expensive or laborious to construct. Based on principle of Rutledge-type feeder, a conventional conical tube, glycerol and Parafilm-M were used to develop a simple in-house feeder device. The blood feeding efficiency of this apparatus was compared to a live blood source, mice, and no significant differences (p = 0.1189) were observed between artificial-fed (51.3% of engorgement) and mice-fed groups (40.6%). Thus, an easy to assemble and cost-effective artificial feeder, designated "Glytube" was developed in this report. This simple and efficient feeding device can be built with common laboratory materials for research on Ae. aegypti.
Sigma-Aldrich
Paraffin wax, mp 53-58 °C (ASTM D 87)
Sigma-Aldrich
Paraffin wax, mp 58-62 °C (ASTM D 87)
Sigma-Aldrich
Paraffin wax, pellets, white, tested according to Ph. Eur.
Sigma-Aldrich
Paraffin wax, tested according to Ph. Eur., high viscosity
Sigma-Aldrich
Paraffin wax, mp ≥65 °C (ASTM D 87)
Sigma-Aldrich
Paraffin wax, pellets, white
Sigma-Aldrich
Paraffin wax, chunks (large), white
Sigma-Aldrich
Paraffin wax, tested according to Ph. Eur., low viscosity
Sigma-Aldrich
Paraffin wax, meets analytical specification of Ph. Eur., pastilles, white