Merck
  • Coexistence of mupirocin and antiseptic resistance in methicillin-resistant Staphylococcus aureus isolates from Korea.
Coexistence of mupirocin and antiseptic resistance in methicillin-resistant Staphylococcus aureus isolates from Korea.
Diagnostic microbiology and infectious disease (2013-01-23)
Hyukmin Lee, Hwansub Lim, Il Kown Bae, Dongeun Yong, Seok Hoon Jeong, Kyungwon Lee, Yunsop Chong
ABSTRACT
In this study, we determined the prevalence of mupirocin and antiseptics resistance in methicillin-resistant Staphylococcus aureus (MRSA) at a secondary- and a tertiary-care hospital in Korea. Mupirocin susceptibility test, ileS gene sequencing, and polymerase chain reaction for mupA, qacA/B, and smr gene were performed with 456 nonduplicated MRSA isolates collected from 2 hospitals in Korea. Genetic relatedness was determined by spa typing and multilocus sequence typing (MLST). The rates of low-level and high-level mupirocin resistance were 7% and 2% in the secondary-care hospital and 17% and 2% in the tertiary-care hospital, respectively. The positive rate of qacA/B and smr gene in mupirocin-resistant MRSA was 65% and 71%, respectively. In spa typing, most mupirocin-resistant MRSA showed genetic relatedness and some of the highly resistant isolates were ST239 and ST5 in MLST analysis. The distribution of low-level and high-level mupirocin resistance and the coexistence of antiseptic resistance in MRSA can result in failure of decolonization in Korea.
Sigma-Aldrich
Lithium mupirocin, ≥95.0% (HPLC)
Sigma-Aldrich
Mupirocin, ≥92% (HPLC), powder