Merck
  • Hepcidin levels predict nonresponsiveness to oral iron therapy in patients with iron deficiency anemia.
Hepcidin levels predict nonresponsiveness to oral iron therapy in patients with iron deficiency anemia.
American journal of hematology (2013-01-22)
David B Bregman, David Morris, Todd A Koch, Andy He, Lawrence T Goodnough
ABSTRACT
Levels of hepcidin, a major regulator of iron homeostasis, may identify patients with iron deficiency anemia (IDA) who will not respond to oral iron therapy. In this study, IDA patients underwent a 14-day trial (run-in) course of ferrous sulfate therapy. Nonresponders (Hgb increase <1 g/dL with 67% compliance rate) were randomized to IV ferric carboxymaltose (FCM; two injections of 750 mg) or further oral iron for 14 days. Screening hepcidin levels were 38.4 versus 11.3 ng/mL, P = 0.0002 in nonresponders versus responders to a trial of oral iron. Hepcidin of > 20 ng/mL, showed sensitivity of 41.3%, specificity of 84.4%, and positive predictive value of 81.6% for predicting nonresponsiveness to oral iron. PPVs for ferritin> 30 ng/mL or transferrin saturation (TSAT)>15% were 59.2 and 55%, respectively. Negative predictive values for hepcidin, ferritin, and TSAT were 46.3, 22.7, and 19.7, respectively. FCM versus oral iron showed Hgb increases of ≥ 1 gm/dL in 65.3% versus 20.8% (P < 0.0001) and Hgb increases of 1.7 ± 1.3 versus 0.6 ± 0.9 g/dL (P = 0.0025), respectively. We conclude that hepcidin predicts nonresponsiveness to oral iron in patients with IDA and is superior to TSAT or ferritin for this purpose. Nonresponse to oral iron therapy does not rule out IDA, since two-thirds of patients subsequently responded to intravenous iron.
Sigma-Aldrich
Iron(II) sulfate heptahydrate, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Iron(II) sulfate heptahydrate, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Iron(II) sulfate heptahydrate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.5-104.5% (manganometric)
Sigma-Aldrich
Iron(II) sulfate heptahydrate, ACS reagent, ≥99.0%
Sigma-Aldrich
Iron(II) sulfate heptahydrate, ReagentPlus®, ≥99%
Sigma-Aldrich
Maltose solution, BioReagent, ~20% in H2O