Merck
  • Prevention of kidney injury following rhabdomyolysis: a systematic review.
Prevention of kidney injury following rhabdomyolysis: a systematic review.
The Annals of pharmacotherapy (2013-01-18)
Elizabeth J Scharman, William G Troutman
ABSTRACT
To conduct a systematic literature review to evaluate evidence-based recommendations for the prevention of rhabdomyolysis-associated acute renal failure (ARF). PubMed (1966-December 2012), International Pharmaceutical Abstracts, Science Citation Index, and Cochrane databases (1970-December 2012) were searched. There were no language restrictions. Studies selected dealt with treatment of rhabdomyolysis (crush syndrome) or prevention of ARF in patients with rhabdomyolysis. Articles excluded did not present original data or described only the management of ARF after it developed. Single case reports were excluded. Extracted data included study type; population; definitions of rhabdomyolysis and ARF; fluid, sodium bicarbonate, and mannitol dosages; and study findings. Twenty-seven studies met the inclusion criteria. No controlled trials compared intravenous fluid administration plus sodium bicarbonate to fluid administration alone. Three concluded that there was no significant difference in the rates of ARF between patients receiving and those not receiving sodium bicarbonate; however, urine alkalinization was not documented. Eight investigations concluded that delayed fluid administration increased the risk of ARF. No controlled study compared volumes of fluid administered or targeted urine output goals. Fluid type, therapy duration, and monitoring parameters varied widely; 4 used a urine output goal in adults of more than 300 mL/h or 300 mL/h or more. No evidence supported a preferred fluid type or that sodium bicarbonate with or without mannitol was superior to fluid therapy alone. Intravenous fluids should be initiated as soon as possible, preferably within the first 6 hours after muscle injury, at a rate that maintains a urine output in adults of 300 mL/h or more for at least the first 24 hours. Sodium bicarbonate should be administered only if necessary to correct systemic acidosis and mannitol only to maintain urine output of 300 mL/h or more despite adequate fluid administration.
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Sodium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Supelco
Sodium bicarbonate concentrate, 0.1 M NaHCO3 in water, eluent concentrate for IC
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium bicarbonate solution, solution (7.5%), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Sodium hydrogencarbonate, −40-+140 mesh, ≥95%