Merck
  • Organocatalytic asymmetric multicomponent reactions of aromatic aldehydes and anilines with β-ketoesters: facile and atom-economical access to chiral tetrahydropyridines.
Organocatalytic asymmetric multicomponent reactions of aromatic aldehydes and anilines with β-ketoesters: facile and atom-economical access to chiral tetrahydropyridines.
Chemical communications (Cambridge, England) (2013-01-12)
Xuejian Li, Yanyan Zhao, Haijun Qu, Zhenjun Mao, Xufeng Lin
ABSTRACT
The first catalytic asymmetric pseudo five-component (AB(2)C(2) type) reaction is reported. A spirocyclic chiral phosphoric acid catalyzed one-pot multicomponent reaction of aromatic aldehydes, anilines and β-ketoesters and afforded highly functionalized enantioenriched tetrahydropyridines with high levels of stereocontrol.
Sigma-Aldrich
Aniline-4-13C, 99 atom % 13C
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Aniline, ReagentPlus®, 99%
Sigma-Aldrich
Aniline-15N, 98 atom % 15N
Sigma-Aldrich
Aniline, ACS reagent, ≥99.5%
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
Supelco
Aniline, analytical standard
Supelco
Aniline solution, certified reference material, 5000 μg/mL in methanol
Sigma-Aldrich
Phosphoric acid, crystalline, ≥99.999% trace metals basis
Sigma-Aldrich
Aniline-1-13C, 99 atom % 13C
Sigma-Aldrich
Phosphoric acid, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Phosphoric acid, BioUltra, ≥85% (T)
Sigma-Aldrich
Phosphoric acid, ≥85 wt. % in H2O, ≥99.999% trace metals basis
Sigma-Aldrich
Aniline hydrochloride, ≥99%
Sigma-Aldrich
Phosphoric acid, BioReagent, suitable for insect cell culture, 85%
Sigma-Aldrich
Phosphoric acid, puriss. p.a., crystallized, ≥99.0% (T)
Sigma-Aldrich
Phosphoric acid, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Phosphoric acid, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Phosphoric acid, puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, 85.0-88.0%
Sigma-Aldrich
Phosphoric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥85%