Merck
  • Cloning and expression of a small heat and salt tolerant protein (Hsp22) from Chaetomium globosum.
Cloning and expression of a small heat and salt tolerant protein (Hsp22) from Chaetomium globosum.
Indian journal of experimental biology (2013-01-12)
Rashmi Aggarwal, Sangeeta Gupta, Sapna Sharma, Sagar Banerjee, Priyanka Singh
ABSTRACT
The present study reports molecular characterization of small heat shock protein gene in Indian isolates of Chaetomium globosum, C. perlucidum, C. reflexum, C. cochlioides and C. cupreum. Six isolates of C. globosum and other species showed a band of 630bp using specific primers. Amplified cDNA product of C. globosum (Cg 1) cloned and sequenced showed 603bp open reading frame encoding 200 amino-acids. The protein sequence had a molecular mass of 22 kDa and was therefore, named Hsp22. BlastX analysis revealed that the gene codes for a protein homologous to previously characterized Hsp22.4 gene from C. globosum (AAR36902.1, XP 001229241.1) and shared 95% identity in amino acid sequence. It also showed varying degree of similarities with small Hsp protein from Neurospora spp. (60%), Myceliophthora sp. (59%), Glomerella sp. (50%), Hypocrea sp. (52%), and Fusarium spp. (51%). This gene was further cloned into pET28a (+) and transformed E. coli BL21 cells were induced by IPTG, and the expressed protein of 30 kDa was analyzed by SDS-PAGE. The IPTG induced transformants displayed significantly greater resistance to NaCl and Na2CO3 stresses.
Product Description
Supelco
Sodium bicarbonate concentrate, 0.1 M NaHCO3 in water, eluent concentrate for IC
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium bicarbonate solution, solution (7.5%), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium hydrogencarbonate, −40-+140 mesh, ≥95%
Sigma-Aldrich
Sodium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C