Merck
  • Transcatheter embolisation of type 1 endoleaks after endovascular aortic aneurysm repair with Onyx: when no other treatment option is feasible.
Transcatheter embolisation of type 1 endoleaks after endovascular aortic aneurysm repair with Onyx: when no other treatment option is feasible.
European journal of vascular and endovascular surgery : the official journal of the European Society for Vascular Surgery (2013-01-02)
J-Y Chun, R Morgan
ABSTRACT
Type 1 endoleaks following endovascular aortic aneurysm repair are associated with poor outcomes and re-intervention is recommended as soon as possible after diagnosis. When standard endovascular or surgical treatment options are unsuitable due to severe co-morbidity or adverse anatomic factors, patients can be treated by transcatheter embolisation of the endoleak itself. We describe six such patients with proximal and distal type 1 endoleaks, who have been successfully treated by transcatheter embolisation with Onyx. The embolisation technique, advantages of using this relatively novel liquid embolic agent and potential pitfalls are discussed.
Sigma-Aldrich
Tantalum, powder, −325 mesh, 99.9% trace metals basis
Sigma-Aldrich
Tantalum, foil, thickness 0.05 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Tantalum, powder, 99.99% trace metals basis
Sigma-Aldrich
Tantalum, foil, thickness 1.0 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Tantalum, foil, thickness 0.025 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Tantalum, foil, thickness 0.25 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Tantalum, foil, thickness 0.5 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Tantalum, wire, diam. 1.0 mm, ≥99.9% trace metals basis