Merck
  • Alkali recovery using PVA/SiO2 cation exchange membranes with different -COOH contents.
Alkali recovery using PVA/SiO2 cation exchange membranes with different -COOH contents.
Journal of hazardous materials (2012-12-29)
Jianwen Hao, Ming Gong, Yonghui Wu, Cuiming Wu, Jingyi Luo, Tongwen Xu
ABSTRACT
By changing -COOH content in poly(acrylic acid-co-methacryloxypropyl trimethoxy silane (poly(AA-co-γ-MPS)), a series of PVA/SiO(2) cation exchange membranes are prepared from sol-gel process of poly(AA-co-γ-MPS) in presence of poly(vinyl alcohol) (PVA). The membranes have the initial decomposition temperature (IDT) values of 236-274 °C. The tensile strength (TS) ranges from 17.4 MPa to 44.4 MPa. The dimensional stability in length (DS-length) is in the range of 10%-25%, and the DS-area is in the range of 21%-56% in 65 °C water. The water content (W(R)) ranges from 61.2% to 81.7%, the ion exchange capacity (IEC) ranges from 1.69 mmol/g to 1.90 mmol/g. Effects of -COOH content on diffusion dialysis (DD) performance also are investigated for their potential applications. The membranes are tested for recovering NaOH from the mixture of NaOH/Na(2)WO(4) at 25 - 45 °C. The dialysis coefficients of NaOH (U(OH)) are in the range of 0.006-0.032 m/h, which are higher than those of the previous membranes (U(OH): 0.0015 m/h, at 25 °C). The selectivity (S) can reach up to 36.2. The DD performances have been correlated with the membrane structure, especially the continuous arrangement of -COOH in poly(AA-co-γ-MPS) chain.
Sigma-Aldrich
3-(Trimethoxysilyl)propyl methacrylate, ≥97%
Sigma-Aldrich
Kuraray Poval ®56-98, Mw ~195,000
Sigma-Aldrich
Mowiol® 28-99, Mw ~145,000
Supelco
Poly(acrylic acid sodium salt), analytical standard, for GPC, 16,000
Sigma-Aldrich
Mowiol® 18-88, Mw ~130,000
Sigma-Aldrich
Poly(acrylic acid), average Mv ~4,000,000
Sigma-Aldrich
Poly(acrylic acid), average Mv ~3,000,000
Sigma-Aldrich
Poly(acrylic acid), average Mv ~1,250,000
Sigma-Aldrich
Poly(acrylic acid sodium salt), average Mw ~5,100 by GPC, powder
Sigma-Aldrich
Poly(acrylic acid), average Mv ~450,000
Supelco
Poly(acrylic acid), analytical standard, average Mn 130,000 (Typical)
Sigma-Aldrich
Poly(acrylic acid)
Sigma-Aldrich
Mowiol® 40-88, average Mw ~205,000 g/mol
Sigma-Aldrich
Poly(acrylic acid sodium salt), average Mw ~2,100
Sigma-Aldrich
3-(Trimethoxysilyl)propyl methacrylate, 98%
Sigma-Aldrich
Mowiol® 4-98, Mw ~27,000
Sigma-Aldrich
Mowiol® 4-88, Mw ~31,000
Sigma-Aldrich
Poly(acrylic acid) solution, average Mw ~2,000, 50 wt. % in H2O, electronic grade
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 13,000-23,000, 98% hydrolyzed
Sigma-Aldrich
Mowiol® 6-98, Mw ~47,000
Sigma-Aldrich
Mowiol® 10-98, Mw ~61,000
Sigma-Aldrich
Poly(vinyl alcohol), Mw 146,000-186,000, 99+% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), Mw 9,000-10,000, 80% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 31,000-50,000, 87-89% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), Mw 85,000-124,000, 99+% hydrolyzed
Sigma-Aldrich
Poly(acrylic acid) solution, average Mw ~100,000, 35 wt. % in H2O
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 85,000-124,000, 87-89% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 146,000-186,000, 87-89% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), Mw 31,000-50,000, 98-99% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), Mw 89,000-98,000, 99+% hydrolyzed