Merck
  • Molecular characterization of high-level mupirocin resistance in Staphylococcus pseudintermedius.
Molecular characterization of high-level mupirocin resistance in Staphylococcus pseudintermedius.
Journal of clinical microbiology (2012-12-28)
Krešimir Matanovic, Eduardo Pérez-Roth, Selma Pintarić, Branka Šeol Martinec
ABSTRACT
The genetic analysis of high-level mupirocin resistance (Hi-Mup(r)) in a Staphylococcus pseudintermedius isolate from a dog is presented. The Hi-Mup(r) ileS2 gene flanked by a novel rearrangement of directly repeated insertion sequence IS257 elements was located, together with the aminoglycoside resistance aacA-aphD determinant, on a conjugative plasmid related to the pSK41/pGO1 family plasmids.
Sigma-Aldrich
Lithium mupirocin, ≥95.0% (HPLC)
Sigma-Aldrich
Mupirocin, ≥92% (HPLC), powder