Merck
Predicting drug release from HPMC/lactose tablets.
International journal of pharmaceutics (2012-12-25)
J Siepmann, Y Karrout, M Gehrke, F K Penz, F Siepmann
ABSTRACT
Three mathematical models were applied to quantify drug release from HPMC/lactose-based matrix tablets loaded with varying amounts of theophylline: (i) a numerical model considering drug diffusion in axial and radial direction in cylinders as well as limited drug solubility effects, (ii) an analytical solution of Fick's second law of diffusion considering axial and radial mass transport in a cylinder, but neglecting limited drug solubility effects, and (iii) a simple early time approximation of the analytical solution, considering only radial mass transport and neglecting axial diffusion as well as limited drug solubility effects. The three models were fitted to experimentally determined drug release kinetics from various types of tablets in 0.1M HCl and phosphate buffer pH 7.4. Interestingly, the agreement between fitted theories and experimental data was similar in all cases. However, the determined system specific model parameters (apparent diffusion coefficients of theophylline in the polymeric matrices) were significantly biased when using simplified theories. Nevertheless, the reliability of theoretical predictions was similar for all three models, since the determined apparent diffusivities are partially "lumped" parameters. Thus, from a practical point of view, very simple equations can be used during product optimization, allowing estimating the effects of formulation parameters on drug release.
Sigma-Aldrich
Methyl cellulose, 27.5-31.5% methoxyl basis
Sigma-Aldrich
Methyl cellulose, tested according to Ph. Eur.
Sigma-Aldrich
Methyl cellulose, medium viscosity, Methoxyl content 27.5-31.5 %
Sigma-Aldrich
Methyl cellulose, viscosity: 4,000 cP
Sigma-Aldrich
Methyl cellulose, viscosity: 15 cP, BioReagent, suitable for cell culture
Sigma-Aldrich
Methyl cellulose, 26.0-33.0% (Methoxy group (dry basis)), meets USP testing specifications, viscosity: 1,500 cP
Sigma-Aldrich
Methyl cellulose, meets USP testing specifications, 26.0-33.0% (methoxyl group, on Dry Basis), viscosity: 400 cP
Sigma-Aldrich
Methyl cellulose, viscosity: 25 cP
Sigma-Aldrich
Methyl cellulose, viscosity: 1,500 cP
Sigma-Aldrich
Methyl cellulose, 27.5-31.5% (Methoxyl content), viscosity: 400 cP
Supelco
Theophylline, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methyl cellulose, viscosity: 15 cP
Sigma-Aldrich
Theophylline, anhydrous, ≥99%, powder
Theophylline, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Methyl cellulose, viscosity 3000-5500 mPa.s, 2 % in H2O(20 °C)
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, viscosity 2,600-5,600 cP, 2 % in H2O(20 °C)(lit.)
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, viscosity 40-60 cP, 2 % in H2O(20 °C)(lit.)
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, average Mn ~90,000
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, viscosity 80-120 cP, 2 % in H2O(20 °C)(lit.)
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, average Mn ~120,000
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, average Mn ~10,000
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, average Mn ~86,000
Sigma-Aldrich
Hypromellose, meets USP testing specifications
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose