Merck
  • Perturbation of thermal unfolding and aggregation of human IgG1 Fc fragment by Hofmeister anions.
Perturbation of thermal unfolding and aggregation of human IgG1 Fc fragment by Hofmeister anions.
Molecular pharmaceutics (2012-12-22)
Jian Zhang-van Enk, Bruce D Mason, Lei Yu, Le Zhang, Wael Hamouda, Gang Huang, Dingjiang Liu, Richard L Remmele, Jifeng Zhang
ABSTRACT
The thermal unfolding and subsequent aggregation of the unglycosylated Fc fragment of a human IgG1 antibody (Fc) were studied in the salt solutions of Na(2)SO(4), KF, KCl and KSCN at pH 4.8 and 7.2 below and at its pI of 7.2, respectively, using differential scanning calorimetry (DSC), far ultraviolet circular dichroism (far-UV CD), size exclusion chromatography (SE-HPLC) and light scattering. First, our experimental results demonstrated that the thermal unfolding of the C(H)2 domain of the Fc was sufficient to induce aggregation. Second, at both pH conditions, the anions (except F(-)) destabilized the C(H)2 domain where the effectiveness of SO(4)(2-) > SCN(-) > Cl(-) > F(-) was more apparent at pH 4.8. In addition, the thermal stability of the C(H)2 domain was less sensitive to the change in salt concentration at pH 7.2 than at pH 4.8. Third, at pH 4.8 when the Fc had a net positive charge, the anions accelerated the aggregation reaction with SO(4)(2-) > SCN(-) > Cl(-) > F(-) in effectiveness. But these anions slowed down the aggregation kinetics at pH 7.2 with similar effectiveness when the Fc was net charge neutral. We hypothesize that the effectiveness of the anion on destabilizing the C(H)2 domain could be attributed to its ability to perturb the free energy for both of the native and unfolded states. The effect of the anions on the kinetics of the aggregation reaction could be interpreted based on the modulation of the electrostatic protein-protein interactions by the anions.
Sigma-Aldrich
Sodium hydrogen sulfate, technical grade
Sigma-Aldrich
Potassium fluoride, ≥99.97% trace metals basis
Sigma-Aldrich
Sodium sulfate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Sodium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium fluoride, BioUltra, ≥99.5% (F)
Sigma-Aldrich
Potassium fluoride, anhydrous, powder, ≥99.9% trace metals basis
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium sulfate, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Sodium sulfate, puriss., meets analytical specification of Ph. Eur., BP, USP, anhydrous, 99.0-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium sulfate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, powder
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, granular
Sigma-Aldrich
Sodium sulfate decahydrate, puriss. p.a., crystallized, ≥99.0% (calc. based on dry substance, T)
Sigma-Aldrich
Sodium bisulfate monohydrate, puriss. p.a., ≥99.0% (T)
Sigma-Aldrich
Sodium sulfate decahydrate, reagent grade, 97%
Sigma-Aldrich
Sodium sulfate decahydrate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium bisulfate monohydrate, ReagentPlus®, 99%