Merck
2D and 3D photoreactive lanthanide MOFs of trans,trans-muconic acid.
Chemical communications (Cambridge, England) (2012-12-20)
Adonis Michaelides, Stavroula Skoulika, Michael G Siskos
ABSTRACT
Three 2D and 3D photoreactive MOFs of trans,trans-muconic acid with Er(3+) and their corresponding isomorphous Y(3+) phases were synthesized and their photoreactivity was studied as a function of the crystal environment.
Sigma-Aldrich
Sorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
Sorbic acid, 99.0-101.0% anhydrous basis
Sigma-Aldrich
trans,trans-Muconic acid, 95%