Merck
  • Crown ether-containing Schiff base as a highly efficient "turn-on" fluorescent sensor for determination and separation of Zn2+ in water.
Crown ether-containing Schiff base as a highly efficient "turn-on" fluorescent sensor for determination and separation of Zn2+ in water.
Dalton transactions (Cambridge, England : 2003) (2012-12-14)
Damir A Safin, Maria G Babashkina, Yann Garcia
ABSTRACT
A highly efficient Zn(2+) "turn-on" fluorescent sensor, [4'-benzo-15-crown-5]N=CH-(2-OH-C(6)H(4)) (HL), has been synthesized. In the presence of Zn(NO(3))(2), displays high selectivity and sensitive enhancement of fluorescence intensity in water with the formation of [ZnL(2)]. Addition of an equimolar amount of KNO(3) to [ZnL(2)] leads to the formation of {[KZnL(2)]NO(3)}(n).
Sigma-Aldrich
Potassium nitrate, 99.99% trace metals basis
Sigma-Aldrich
Potassium nitrate, 99.999% trace metals basis
Sigma-Aldrich
Potassium nitrate, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium nitrate, BioReagent, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Potassium nitrate, ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium nitrate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Potassium nitrate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Potassium nitrate, puriss., meets analytical specification of Ph Eur, BP, USP, FCC, E252, 99.0-100.5%
Supelco
Mettler-Toledo Calibration substance ME 51143095, Potassium nitrate, traceable to primary standards (LGC)
Sigma-Aldrich
15-Crown-5, 98%