Merck
  • Ampicillin alone and in combination with riboflavin modulates Staphylococcus aureus infection induced septic arthritis in mice.
Ampicillin alone and in combination with riboflavin modulates Staphylococcus aureus infection induced septic arthritis in mice.
Indian journal of experimental biology (2012-12-12)
Pinky Mal, Deboshree Ghosh, Debasish Bandyopadhyay, Kallol Dutta, Biswadev Bishayi
ABSTRACT
Effects of ampicillin (Amp) in combination with riboflavin on septic arthritis in mice infected with Staphylococcus aureus have been reported. Ampicillin was given at 100 mg/kg after 24 h of infection, followed by riboflavin (Ribo) at 20 mg/kg body wt, after 2 h of Amp treatment. Mice were sacrificed at 3, 9, 15 days post infection (dpi). Combined treatment of infected mice with ampicillin and riboflavin eradicated the bacteria from blood, spleen and synovial tissue and showed a significant gross reduction in arthritis, reduced serum levels of TNF-alpha and IFN-gamma. S. aureus infected mice exhibited higher synovial TNF-alpha and IL-6, which was also reduced by ampicillin and riboflavin treatment. S. aureus infected mice showed a disturbed antioxidant status measured in terms of cellular anti-oxidants like reduced glutathione and anti-oxidant enzymes such as superoxide dismutase and catalase and were ameliorated when the animals were co-treated with ampicillin along with riboflavin. Results of the study showed that combined treatment with anti-oxidant and antibiotic may protect from staphylococcal arthritis and may ameliorate oxidative stress caused by S. aureus infection.
Product Description
Supelco
Ampicillin trihydrate, VETRANAL®, analytical standard
Sigma-Aldrich
Ampicillin trihydrate, 900-1050 μg/mg anhydrous basis (HPLC)
Sigma-Aldrich
Ampicillin, meets USP testing specifications
Sigma-Aldrich
Ampicillin sodium salt
Sigma-Aldrich
Ampicillin sodium salt, powder or crystals, BioReagent, suitable for cell culture
Sigma-Aldrich
Ampicillin sodium salt, BioXtra, suitable for cell culture