Merck
Occurrence and dynamics of micropollutants in a karst aquifer.
Environmental pollution (Barking, Essex : 1987) (2012-12-04)
Barbara Morasch
ABSTRACT
Karst systems represent important yet vulnerable drinking water resources. A wide spectrum of pollutants may be released into karst groundwater from agriculture, livestock farming, private households, and industry. This work provides an overview on the occurrence and dynamics of micropollutants in a karst system of the Swiss Jura. Ten months of intensive monitoring for micropollutants confirmed that the swallow hole draining an agricultural plain was the main entry path for pesticides into the karst system and the two connected springs. Elevated fungicide concentrations in winter and occasional quantification of pharmaceuticals suggested wood- or façade treatment and domestic sewer as additional sources of contamination. A continuous atrazine signal in the low ng/L range might affect the autochthonous endokarst microbial community and represents a potential risk for the human population through karst groundwater.
Sigma-Aldrich
Drierite, with indicator, 10-20 mesh
Sigma-Aldrich
Drierite, with indicator, 6 mesh
Sigma-Aldrich
Drierite, without indicator, 8 mesh
Sigma-Aldrich
Drierite, with indicator, 8 mesh
Sigma-Aldrich
Drierite, with indicator, 4 mesh
Sigma-Aldrich
Drierite, without indicator, 4 mesh
Sigma-Aldrich
Calcium sulfate hemihydrate, purum, ≥97.0%
Sigma-Aldrich
Calcium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Calcium sulfate dihydrate, puriss., meets analytical specification of NF, E 516, 99.0-101.0% (based on anhydrous substance)
Sigma-Aldrich
Calcium sulfate dihydrate, ReagentPlus®, ≥99%
Sigma-Aldrich
Calcium sulfate dihydrate, ACS reagent, 98%
Sigma-Aldrich
Calcium sulfate, −325 mesh, 99%