Merck
  • Ursolic acid, a potential PPARγ agonist, suppresses ovalbumin-induced airway inflammation and Penh by down-regulating IL-5, IL-13, and IL-17 in a mouse model of allergic asthma.
Ursolic acid, a potential PPARγ agonist, suppresses ovalbumin-induced airway inflammation and Penh by down-regulating IL-5, IL-13, and IL-17 in a mouse model of allergic asthma.
European journal of pharmacology (2012-12-04)
Seung-Hyung Kim, Jung-Hee Hong, Young-Cheol Lee
ABSTRACT
Allergic asthma is a chronic airway disorder characterized by airway hyperresponsiveness to allergens, chronic airway inflammation, airway edema, increased mucus secretion, excess production of Th2 cytokines, and eosinophil accumulation in the lungs. Ursolic acid is known for its pharmacological effects, such as its anti-tumor, anti-inflammatory and antimicrobial activities. To investigate the anti-asthmatic effects and mechanism of ursolic acid, we studied the development of pulmonary eosinophilic inflammation and enhanced pause (Penh) in a mouse model of allergic asthma. In this study, BALB/c mice were systemically sensitized to ovalbumin followed by intratracheal, intraperitoneal, and aerosol allergen challenges. We investigated the effect of ursolic acid and Cyclosporin A (CsA) on Penh, pulmonary eosinophilic infiltration, various immune cell phenotypes, Th2 cytokines, IL-17 production, and ovalbumin specific IgE production in a mouse model of asthma. In BALB/c mice, ursolic acid had suppressed eosinophil infiltration, allergic airway inflammation, and Penh, which occurred by suppressing the production of IL-5, IL-13, IL-17, and ovalbumin-specific IgE by blocking the GATA-3 and STAT6 pathways. Our data suggest the therapeutic mechanism of ursolic acid in asthma is based on reductions of Th2 cytokines (IL-5 and IL-13), ovalbumin-specific IgE production, and eosinophil infiltration via the Th2-GATA-3, STAT6, and IL-17-NF-κB pathways.
Product Description
Supelco
Ursolic acid, analytical standard
Sigma-Aldrich
Ursolic acid, ≥90%
Ursolic acid, primary reference standard