Merck
  • Multiresidue determination of 375 organic contaminants including pesticides, polychlorinated biphenyls and polyaromatic hydrocarbons in fruits and vegetables by gas chromatography-triple quadrupole mass spectrometry with introduction of semi-quantification approach.
Multiresidue determination of 375 organic contaminants including pesticides, polychlorinated biphenyls and polyaromatic hydrocarbons in fruits and vegetables by gas chromatography-triple quadrupole mass spectrometry with introduction of semi-quantification approach.
Journal of chromatography. A (2012-11-28)
Kaushik Banerjee, Sagar Utture, Soma Dasgupta, Chandrasekar Kandaswamy, Saswati Pradhan, Sunil Kulkarni, Pandurang Adsule
ABSTRACT
A residue analysis method for the simultaneous estimation of 349 pesticides, 11 PCBs and 15 PAHs extracted from grape, pomegranate, okra, tomato and onion matrices, was established by using a gas chromatograph coupled to an electron impact ionization triple quadrupole mass spectrometer (GC-EI-MS/MS). The samples were extracted by ethyl acetate and cleaned by dispersive solid phase extraction with PSA and/or GCB/C(18) by the methods reported earlier. The GC-EI-MS/MS parameters were optimized for analysis of all the 375 compounds within a 40 min run time with limit of quantification for most of the compounds at <10 μg/L, which is well below their respective European Union-Maximum Residue Levels. The coefficient of determination (r(2)) was >0.99 within the calibration linearity range of <5-250 ng/mL for compounds with LOQs<5 ng/mL. While for the compounds with LOQs within 5-10 μg/kg, the lowest calibration level was 5 and 10 μg/kg as applicable. The recoveries at 10, 25 and 50 ng/mL were within 70-110% (n=6) with associated RSDs<20% indicating satisfactory precision. The information generated from the single laboratory validation was further utilized for building a semi-quantitative approach. The accuracies in quantification obtained via individual calibration standards vis-à-vis semi-quantification approach were comparable. For incurred samples, the concentrations estimated by the semi-quantification approach were within ±10% of the values obtained by direct quantification. This approach complements the existing GC-EI-MS/MS methods by offering targeted screening and quantification capabilities.
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
Ethyl acetate, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Supelco
Ethyl acetate, analytical standard
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Ethyl acetate, biotech. grade, ≥99.8%
Sigma-Aldrich
Ethyl acetate, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99.5% (GC)