Merck
  • Multiplex bead suspension array for screening Neisseria gonorrhoeae antibiotic resistance genetic determinants in noncultured clinical samples.
Multiplex bead suspension array for screening Neisseria gonorrhoeae antibiotic resistance genetic determinants in noncultured clinical samples.
The Journal of molecular diagnostics : JMD (2012-11-20)
Sergey Balashov, Eli Mordechai, Martin E Adelson, Scott E Gygax
ABSTRACT
The increasing threat of antibiotic-resistant Neisseria gonorrhoeae highlights the need for new diagnostic options. A high-throughput multiplex bead suspension array assay was developed for profiling 29 N. gonorrhoeae genomic mutations and 2 plasmid genes conferring resistance to 6 antimicrobial agents: penicillin, ciprofloxacin, cefixime, tetracycline, azithromycin, and spectinomycin. The three steps of this assay include amplification of 12 N. gonorrhoeae chromosomal and plasmid loci, multiplex allele-specific primer extension reaction, and multiplex bead suspension array detection. Antibiotic resistance genetic determinants were identified successfully in 239 cervicovaginal N. gonorrhoeae-positive noncultured swab samples. This molecular assay can be used for detection of gonococci in clinical specimens, molecular typing, mutation profiling, and predictive assessment of N. gonorrhoeae susceptibility to antibiotics without the need for culture.
Sigma-Aldrich
Spectinomycin dihydrochloride pentahydrate, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Spectinomycin dihydrochloride pentahydrate, potency: ≥603 μg per mg
Sigma-Aldrich
Spectinomycin dihydrochloride pentahydrate, powder, γ-irradiated
Supelco
Spectinomycin dihydrochloride pentahydrate, VETRANAL®, analytical standard