Merck
HPLC analysis and cytotoxic activity of Vernonia cinerea.
Natural product communications (2012-11-20)
Mom Khay, Phirom Toeng, Valérie Mahiou-Leddet, Fathi Mabrouki, Kim Sothea, Evelyne Ollivier, Riad Elias, Sok-Siya Bun
ABSTRACT
The extracts of five Cambodian medicinal plants (Aganosma marginata, Dracaena cambodiana, Harrisonia perforata, Hymenodictyon excelsum and Vernonia cinerea) were evaluated in vitro for their cytotoxic activity against HT29 colon adenocarcinoma cells and HepG2 hepatoma cells, using the MTT assay. Among these five plants, Vernonia cinerea displayed potent cytotoxicity. One main sesquiterpene lactone, 8alpha-tigloyloxy-hirsutinolide-13-O-acetate was isolated from the whole plant of V. cinerea. This compound was active against both cancer cell lines (IC50 = 3.50 microM for HT29 and IC50 = 4.27 microM for HepG2). To quantify this compound in the plant, an analytical high-performance liquid chromatography (HPLC) method was developed and validated.
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Supelco
Dichloromethane, Selectophore, ≥99.5%
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Supelco
Dichloromethane, analytical standard
Sigma-Aldrich
Dichloromethane, puriss., meets analytical specification of Ph. Eur., NF, ≥99% (GC)
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, biotech. grade, 99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, puriss. p.a., ACS reagent, reag. ISO, ≥99.9% (GC)
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.8%, contains amylene as stabilizer
Sigma-Aldrich
Dichloromethane, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 50-150 ppm amylene as stabilizer