Merck
  • Sonodynamic and sonocatalytic damage of BSA molecules by Cresol Red, Cresol Red-DA and Cresol Red-DA-Fe under ultrasonic irradiation.
Sonodynamic and sonocatalytic damage of BSA molecules by Cresol Red, Cresol Red-DA and Cresol Red-DA-Fe under ultrasonic irradiation.
Ultrasonics sonochemistry (2012-11-17)
Mingming Zou, Ying Li, Jun Wang, Qi Wang, Jingqun Gao, Qi Yang, Ping Fan
ABSTRACT
The interaction of Cresol Red derivatives (Cresol Red (o-Cresolsulfonphthalein), Cresol Red-DA (3,3'-Bis [N,N-di (carboxymethyl) aminomethyl]-o-cresolsulfonphthalein) and Cresol Red-DA-Fe(III) (3,3'-Bis [N,N-di (carboxymethyl) aminomethyl]-o-cresolsulfonphthalein-Ferrous(III)) with bovine serum albumin (BSA) were studied by the combination of ultraviolet spectroscopy, circular dichroism (CD) spectroscopy, fluorescence spectroscopy and synchronous fluorescence spectroscopy. On that basis, the sonodynamic and sonocatalytic damages of Cresol Red derivatives to BSA under ultrasonic irradiation were also investigated by means of corresponding spectrum technology. Meanwhile, some influenced factors such as ultrasonic irradiation time, Cresol Red derivatives concentration and ionic strength on the damage degree of BSA molecules were also reviewed. In addition, the binding site and damage site of BSA molecules were estimated by synchronous fluorescence spectra. Finally, the results of oxidation-extraction photometry (OEP) using several reactive oxygen species (ROS) scavengers indicated that the damage of BSA molecules is mainly due to the generation of ROS. Perhaps, this paper may offer some important subjects for broadening the application of Cresol Red derivatives in sonodynamic therapy (SDT) and sonocatalytic therapy (SCT) technologies for tumor treatment.
Sigma-Aldrich
Phenol Red, ACS reagent
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
1 Estimated to ship on March 10, 2025
$76.60
25 g
1 Estimated to ship on March 10, 2025
$224.00
Sigma-Aldrich
Cresol red, indicator grade, Dye content 95 %
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
1 Estimated to ship on March 10, 2025
$82.70
25 g
1 Estimated to ship on March 10, 2025
$287.00
Sigma-Aldrich
Cresol Red sodium salt, indicator grade
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
1 Estimated to ship on March 10, 2025
$82.70
25 g
1 Estimated to ship on March 10, 2025
$287.00
Sigma-Aldrich
Phenol red solution, 0.5%, liquid, sterile-filtered, BioReagent, suitable for cell culture
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
1 Estimated to ship on March 10, 2025
$82.70
25 g
1 Estimated to ship on March 10, 2025
$287.00
Sigma-Aldrich
Phenol Red, powder, BioReagent, suitable for cell culture
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
1 Estimated to ship on March 10, 2025
$82.70
25 g
1 Estimated to ship on March 10, 2025
$287.00