Merck
Bending rigid molecular rods: formation of oligoproline macrocycles.
Chemistry (Weinheim an der Bergstrasse, Germany) (2012-11-06)
Conor C G Scully, Vishal Rai, Gennadiy Poda, Serge Zaretsky, Darcy C Burns, R Scott Houliston, Tiantong Lou, Andrei K Yudin
ABSTRACT
Bent but not broken: cyclic oligoprolines are accessed in a reaction that effectively bends rigid oligoproline peptides (see scheme; TBDMS=tert-butyldimethylsilyl). The stitching is accomplished during macrocyclization enabled by aziridine aldehydes and isocyanides. Molecular modeling studies suggest that electrostatic attraction between the termini of the linear peptide is pivotal for macrocyclization. The macrocycles were studied by circular dichroism with a polyproline II structure being observed in larger macrocycles.
Sigma-Aldrich
Poly-L-proline, mol wt 1,000-10,000
Sigma-Aldrich
Poly-L-proline, mol wt >30,000