Merck
  • Characterization of the interaction between chitosan and inorganic sodium phosphates by means of rheological and optical microscopy studies.
Characterization of the interaction between chitosan and inorganic sodium phosphates by means of rheological and optical microscopy studies.
Carbohydrate polymers (2012-11-06)
Luca Casettari, Marco Cespi, Giovanni Filippo Palmieri, Giulia Bonacucina
ABSTRACT
The physicochemical and rheological properties of chitosan and two different inorganic sodium phosphate dispersions (NaH(2)PO(4) and Na(3)PO(4)) were investigated in order to elucidate the role of different factors, such as ratios between polymer and sodium inorganic phosphates, different pHs and storage stability, on the gelling properties of chitosan. This was deemed opportune since physico-chemical characterizations of chitosan in the literature often appear incomplete and questionable. We also compared the elastic modulus values of the different chitosan/inorganic phosphate systems and examined their behaviour through optical microscopy analyses. The most efficient formulations that showed a thermogelling capacity with a significant gel transition behaviour after 24h were the NaH(2)PO(4)/chitosan and Na(3)PO(4)/chitosan systems at ratio 2 and pH 7.0. These results confirmed the importance of the pH value and ratio selection for the final systems.
Sigma-Aldrich
Sodium phosphate dibasic solution, BioUltra, 0.5 M in H2O
SKU
Pack Size
Availability
Price
Quantity
Supelco
Phosphate Standard for IC, TraceCERT®, 1000 mg/L phosphate in water (nominal concentration)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium phosphate tribasic dodecahydrate, ≥98%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium phosphate tribasic dodecahydrate, BioXtra, ≥98.0% (titration)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium phosphate tribasic dodecahydrate, puriss. p.a., ACS reagent, ≥98.0% (T)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium phosphate tribasic dodecahydrate, ACS reagent, ≥98%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium phosphate dibasic, BioXtra, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium phosphate dibasic, for molecular biology, ≥98.5% (titration)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium phosphate monobasic, meets USP testing specifications, anhydrous
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium phosphate monobasic, ReagentPlus®, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium phosphate dibasic, ReagentPlus®, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium phosphate monobasic, BioXtra, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium phosphate monobasic, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0% (titration)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium phosphate dibasic heptahydrate, meets USP testing specifications
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium phosphate monobasic, BioReagent, for molecular biology, anhydrous, ≥98%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium phosphate dibasic, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, BioXtra, ≥99.0% (T)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium phosphate dibasic, purum p.a., anhydrous, ≥98.0% (T)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium phosphate dibasic, BioUltra, for molecular biology, ≥99.5% (T)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium phosphate monobasic, purum p.a., anhydrous, ≥99.0% (T)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium phosphate, 96%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium phosphate dibasic heptahydrate, ≥99.99% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, tested according to Ph. Eur.
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium phosphate monobasic solution, BioUltra, 5 M in H2O
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium phosphate dibasic, 99.95% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, puriss. p.a., crystallized, ≥99.0% (T)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium phosphate dibasic, ACS reagent, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium phosphate dibasic, puriss. p.a., ACS reagent, anhydrous, ≥99.0% (T)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium phosphate dibasic heptahydrate, ACS reagent, 98.0-102.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium phosphate dibasic, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%, free-flowing, Redi-Dri