Merck
  • Data analysis considerations for detecting copy number changes in formalin-fixed, paraffin-embedded tissues.
Data analysis considerations for detecting copy number changes in formalin-fixed, paraffin-embedded tissues.
Cold Spring Harbor protocols (2012-11-03)
Sharoni Jacobs
ABSTRACT
The Whole Genome Sampling Analysis (WGSA) assay in combination with Affymetrix GeneChip Mapping Arrays is used for copy number analysis of high-quality DNA samples (i.e., samples that have been collected from blood, fresh or frozen tissue, or cell lines). Formalin-fixed, paraffin-embedded (FFPE) samples, however, represent the most prevalent form of archived clinical samples, but they provide additional challenges for molecular assays. FFPE processing usually results in the degradation of FFPE DNA and in the contamination and chemical modification of these DNA samples. In this article, we describe the steps needed to obtain reliable copy number predictions from degraded and contaminated FFPE samples.
Sigma-Aldrich
Paraffin wax, tested according to Ph. Eur., low viscosity
Sigma-Aldrich
Paraffin wax, tested according to Ph. Eur., high viscosity
Sigma-Aldrich
Paraffin wax, chunks (large), white
Sigma-Aldrich
Paraffin wax, pellets, white, tested according to Ph. Eur.
Sigma-Aldrich
Paraffin wax, meets analytical specification of Ph. Eur., pastilles, white
Sigma-Aldrich
Paraffin wax, mp ≥65 °C (ASTM D 87)
Sigma-Aldrich
Paraffin wax, mp 53-58 °C (ASTM D 87)
Sigma-Aldrich
Paraffin wax, mp 58-62 °C (ASTM D 87)
Sigma-Aldrich
Paraffin wax, pellets, white