Merck
Aminoglycoside antibiotics in the 21st century.
ACS chemical biology (2012-11-01)
Bernd Becker, Matthew A Cooper
ABSTRACT
Aminoglycoside antibiotics were among the first antibiotics discovered and used clinically. Although they have never completely fallen out of favor, their importance has waned due to the emergence of other broad-spectrum antibiotics with fewer side effects. Today, with the dramatically increasing rate of infections caused by multidrug-resistant bacteria, focus has returned to aminoglycoside antibiotics as one of the few remaining treatment options, particularly for Gram-negative pathogens. Although the mechanisms of resistance are reasonably well understood, our knowledge about the mode of action of aminoglycosides is still far from comprehensive. In the face of emerging bacterial infections that are virtually untreatable, it is time to have a fresh look at this old class to reinvigorate the struggle against multidrug-resistant pathogens.
Sigma-Aldrich
Hygromycin B solution from Streptomyces hygroscopicus, ≥60% (HPLC), 45-60 mg/mL in H2O, γ-irradiated
Sigma-Aldrich
Hygromycin B solution from Streptomyces hygroscopicus, ≥60% (HPLC), 45-60 mg/mL in H2O
Sigma-Aldrich
Hygromycin B from Streptomyces hygroscopicus, suitable for plant cell culture, BioReagent, ≥60% (HPLC), lyophilized powder
Sigma-Aldrich
Hygromycin B from Streptomyces hygroscopicus, powder, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Hygromycin B from Streptomyces hygroscopicus, lyophilized powder