- Synthesis, and in vitro enzymatic and antiviral evaluation of d4T polyphosphate derivatives as chain terminators.
Synthesis, and in vitro enzymatic and antiviral evaluation of d4T polyphosphate derivatives as chain terminators.
Chemistry & biodiversity (2012-10-20)
Shiqiong Yang, Christophe Pannecouque, Piet Herdewijn
PMID23081918
ABSTRACT
A series of d4T di- or triphosphate derivatives have been synthesized and evaluated as effective substrates for HIV-1 RT, and also tested for their in vitro anti-HIV activity. The steady-state kinetic study of compounds 1-4 in an enzymatic incorporation assay by HIV-1 RT follows Michaelis-Menten profile. In addition, compounds 2-4 are able to inhibit HIV-1 replication to the same extent as d4T and d4TMP in MT-4 cells, as well as in CEM/0 cells and CEM/TK(-) cells. The data suggests that these d4T polyphosphate derivatives are hydrolyzed to d4T and rephosphorylated to d4TTP before exerting their antiviral activity.