Merck
  • Sequential extraction method for determination of Fe(II/III) and U(IV/VI) in suspensions of iron-bearing phyllosilicates and uranium.
Sequential extraction method for determination of Fe(II/III) and U(IV/VI) in suspensions of iron-bearing phyllosilicates and uranium.
Environmental science & technology (2012-10-19)
Fubo Luan, William D Burgos
ABSTRACT
Iron-bearing phyllosilicates strongly influence the redox state and mobility of uranium because of their limited hydraulic conductivity, high specific surface area, and redox reactivity. Standard extraction procedures cannot be accurately applied for the determination of clay-Fe(II/III) and U(IV/VI) in clay mineral-U suspensions such that advanced spectroscopic techniques are required. Instead, we developed and validated a sequential extraction method for determination of clay-Fe(II/III) and U(IV/VI) in clay-U suspensions. In our so-called "H(3)PO(4)-HF-H(2)SO(4) sequential extraction" method, H(3)PO(4)-H(2)SO(4) is used first to solubilize and remove U, and the remaining clay pellet is subject to HF-H(2)SO(4) digestion. Physical separation of U and clay eliminates valence cycling between U(IV/VI) and clay-Fe(II/III) that otherwise occurred in the extraction solutions and caused analytical discrepancies. We further developed an "automated anoxic KPA" method to measure soluble U(VI) and total U (calculate U(IV) by difference) and modified the conventional HF-H(2)SO(4) digestion method to eliminate a series of time-consuming weighing steps. We measured the kinetics of uraninite oxidation by nontronite using this sequential extraction method and anoxic KPA method and measured a stoichiometric ratio of 2.19 ± 0.05 mol clay-Fe(II) produced per mol U(VI) produced (theoretical value of 2.0). We found that we were able to recover 98.0-98.5% of the clay Fe and 98.1-98.5% of the U through the sequential extractions. Compared to the theoretical stoichiometric ratio of 2.0, the parallel extractions of 0.5 M HCl for clay-Fe(II) and 1 M NaHCO(3) for U(VI) leached two-times more Fe(II) than U(VI). The parallel extractions of HF-H(2)SO(4) for clay Fe(II) and 1 M NaHCO(3) for U(VI) leached six-times more Fe(II) than U(VI).
Sigma-Aldrich
Hydrofluoric acid, ACS reagent, 48%
Sigma-Aldrich
Hydrofluoric acid, 48 wt. % in H2O, ≥99.99% trace metals basis