Merck
  • Conductive photoswitchable vanadium oxide nanopaper based on bacterial cellulose.
Conductive photoswitchable vanadium oxide nanopaper based on bacterial cellulose.
ChemSusChem (2012-10-13)
Junkal Gutierrez, Susana C M Fernandes, Iñaki Mondragon, Agnieszka Tercjak
ABSTRACT
A bacterial cellulose mat was used as a template for the fabrication of conductive photoswitchable hybrid nanopaper by the incorporation of sol-gel synthesized vanadium nanoparticles. The resulting nanopaper, prepared through a green pathway, was able to photoinduce a reversible color change. Conductive properties at the nano- and macroscales were confirmed by electrostatic force microscopy and semiconductor analysis measurements, respectively.
Sigma-Aldrich
Vanadium(V) oxide, 99.9% trace metals basis
Sigma-Aldrich
Vanadium(V) oxide, ≥99.6% trace metals basis
Sigma-Aldrich
Vanadium(V) oxide, ≥98%