Merck
  • Determination of the tolerable upper intake level of leucine in acute dietary studies in young men.
Determination of the tolerable upper intake level of leucine in acute dietary studies in young men.
The American journal of clinical nutrition (2012-09-07)
Rajavel Elango, Karen Chapman, Mahroukh Rafii, Ronald O Ball, Paul B Pencharz
ABSTRACT
Leucine has been suggested to improve athletic performance. Therefore, the branched-chain amino acids (BCAAs), especially leucine, are popular as dietary supplements in strength-training athletes; however, the intake of leucine in excess of requirements raises concerns regarding adverse effects. Currently, the tolerable upper intake level (UL) for leucine is unknown. The objective of the current study was to determine the UL for leucine in adult men under acute dietary conditions. Five healthy adults (20-35 y) each received graded stepwise increases in leucine intakes of 50, 150, 250, 500, 750, 1000, and 1250 mg · kg⁻¹ · d⁻¹, which corresponded to the Estimated Average Requirement (EAR) and the EAR ×3, ×5, ×10, ×15, ×20, and ×25 in a total of 29 studies. The UL of leucine was identified by the measurement of plasma and urinary biochemical variables and changes in leucine oxidation by using l-[1-¹³C]-leucine. A significant increase in blood ammonia concentrations above normal values, plasma leucine concentrations, and urinary leucine excretion were observed with leucine intakes >500 mg · kg⁻¹ · d⁻¹. The oxidation of l-[1-¹³C]-leucine expressed as label tracer oxidation in breath (F¹³CO₂), leucine oxidation, and α-ketoisocaproic acid (KIC) oxidation led to different results: a plateau in F¹³CO₂ observed after 500 mg · kg⁻¹ · d⁻¹, no clear plateau observed in leucine oxidation, and KIC oxidation appearing to plateau after 750 mg · kg⁻¹ · d⁻¹. On the basis of plasma and urinary variables, the UL for leucine in healthy adult men can be suggested at 500 mg · kg⁻¹ · d⁻¹ or ~35 g/d as a cautious estimate under acute dietary conditions.
Sigma-Aldrich
4-Methyl-2-oxovaleric acid, ≥98.0% (T)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium 4-methyl-2-oxovalerate, leucine metabolite