Merck
  • Diethyldithiocarbamate complex with copper: the mechanism of action in cancer cells.
Diethyldithiocarbamate complex with copper: the mechanism of action in cancer cells.
Mini reviews in medicinal chemistry (2012-08-31)
Zdenek Skrott, Boris Cvek
ABSTRACT
The idea of "repurposing" of existing drugs provides an effective way to develop and identify new therapies. Disulfiram (Antabuse), a drug commonly used for the treatment of alcoholism, shows promising anticancer activity in both preclinical and clinical studies. In the human body, disulfiram is rapidly converted to its reduced metabolite, diethyldithiocarbamate. If copper ions are available, a bis(diethyldithiocarbamate)-copper(II) complex is formed. Disulfiram's selective anticancer activity is attributed to the copper(II) complex's ability to inhibit the cellular proteasome. It is assumed that the complex inhibits the proteasome by a mechanism that is distinct to the clinically used drug bortezomib, targeting the 19S rather than the 20S proteasome. This difference could be explained by inhibition of the JAMM domain of the POH1 subunit within the lid of the 19S proteasome.
Sigma-Aldrich
Sodium diethyldithiocarbamate trihydrate, ACS reagent
Sigma-Aldrich
Sodium diethyldithiocarbamate trihydrate
Supelco
Sodium diethyldithiocarbamate trihydrate, ACS reagent, for the determination and separation of heavy metals, ≥97%
Sigma-Aldrich
Ammonium diethyldithiocarbamate