Merck
  • Evaluation of the functional importance of organic cation transporters on the ocular disposition of its intravitreally injected substrate in rabbits.
Evaluation of the functional importance of organic cation transporters on the ocular disposition of its intravitreally injected substrate in rabbits.
Current eye research (2012-08-25)
Jayabalan Nirmal, Thirumurthy Velpandian, Sundararajan Baskar Singh, Nihar Ranjan Biswas, Rajvardhan Azad, Vasantha Thavaraj, Gaurav Mittal, Aseem Bhatnagar, Supriyo Ghose
ABSTRACT
To evaluate the functional role of organic cation transporters (OCT) and ocular tissue distribution of intravitreally injected OCT substrate tetraethylammonium (TEA) in presence of OCT blocker (quinidine). New Zealand albino rabbits of either sex were used. Intravitreal quinidine pretreatment was made 30 min before the administration of TEA. Modulation of vitreous and ocular tissue kinetics of OCT substrate was evaluated with or without blocker pretreatment. Gamma scintigraphy was also performed to visualize the vitreous residence of (99m)Tc-labelled TEA in the presence and absence of blocker. Intravitreally injected quinidine did not significantly alter the ocular disposition of TEA. TEA showed less significant posterior elimination kinetics and slow anterior elimination which resulted in longer residence time of TEA in eye after intravitreal administration. Intravitreally injected OCT substrates may follow an anterior elimination pathway and prolonged residence time in vitreous humor. The present study shows that OCT may not be active from vitreous-to-blood route in the blood-retinal barrier.
Sigma-Aldrich
Tetraethylammonium chloride hydrate
Sigma-Aldrich
Tetraethylammonium iodide, 98%
Sigma-Aldrich
Tetraethylammonium chloride, BioUltra, Molecular Biology, ≥99.0% (AT)
Sigma-Aldrich
Tetraethylammonium hydroxide solution, ~25% in methanol (~1.5 M)
Sigma-Aldrich
Tetraethylammonium bromide, reagent grade, 98%
Sigma-Aldrich
Tetraethylammonium hydroxide solution, 20 wt. % in H2O
Sigma-Aldrich
Tetraethylammonium hydroxide solution, 35 wt. % in H2O
Sigma-Aldrich
Tetraethylammonium chloride, ≥98% (titration)
Supelco
Tetraethylammonium hydroxide solution, ~1.0 M (CH3CH2)4NOH in H2O, electrochemical grade
Supelco
Tetraethylammonium chloride, for electrochemical analysis, ≥99.0%
Sigma-Aldrich
Tetraethylammonium bromide, ReagentPlus®, 99%