Merck
  • Chemical composition and antimicrobial activity of essential oil of Heracleum rigens.
Chemical composition and antimicrobial activity of essential oil of Heracleum rigens.
Natural product communications (2012-08-23)
Nataraj Jagannath, Hanumanthaiah Ramakrishnaiah, Venkatarangaiah Krishna, Prameela Javarayi Gowda
ABSTRACT
The essential oil was extracted from the seeds of Heracleum rigens by hydrodistillation and a total of twenty compounds accounting for 98.5% of the total oil composition were identified. Physicochemical properties and chemical composition of the oil was determined by a combination GC/FID and GC/MS analysis. The major compounds identified were bornyl acetate (51.2%), alpha-pinene (22.6%), limonene (9.62%), octyl acetate (3.94%), rho-cymene (2.85%) and gamma-terpinene (1.93%). The antimicrobial activity of the oil was screened by the disc diffusion method against nine pathogenic bacterial strains. Maximum antimicrobial activity was noted against Klebsiella pneumonia and Bacillus subtillis. This investigation corroborates the traditional claim of H. rigens as an effective antimicrobial agent.
Sigma-Aldrich
α-Pinene, 98%
Pricing and availability is not currently available.
α-Pinene, primary reference standard
Pricing and availability is not currently available.
Sigma-Aldrich
α-Terpinene, ≥95.0% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
γ-Terpinene, 97%
Pricing and availability is not currently available.
Sigma-Aldrich
γ-Terpinene, ≥95%, FG
Pricing and availability is not currently available.
Sigma-Aldrich
α-Terpinene, ≥89%, FCC, FG
Pricing and availability is not currently available.
Supelco
γ-Terpinene, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
γ-Terpinene, natural, 95%, FG
Pricing and availability is not currently available.
Sigma-Aldrich
γ-Terpinene, ≥95.0% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
α-Terpinene, 85%
Pricing and availability is not currently available.