Merck
Construction and testing of coin cells of lithium ion batteries.
Journal of visualized experiments : JoVE (2012-08-17)
Archana Kayyar, Jiajia Huang, Mojtaba Samiee, Jian Luo
ABSTRACT
Rechargeable lithium ion batteries have wide applications in electronics, where customers always demand more capacity and longer lifetime. Lithium ion batteries have also been considered to be used in electric and hybrid vehicles or even electrical grid stabilization systems. All these applications simulate a dramatic increase in the research and development of battery materials, including new materials, doping, nanostructuring, coatings or surface modifications and novel binders. Consequently, an increasing number of physicists, chemists and materials scientists have recently ventured into this area. Coin cells are widely used in research laboratories to test new battery materials; even for the research and development that target large-scale and high-power applications, small coin cells are often used to test the capacities and rate capabilities of new materials in the initial stage. In 2010, we started a National Science Foundation (NSF) sponsored research project to investigate the surface adsorption and disordering in battery materials (grant no. DMR-1006515). In the initial stage of this project, we have struggled to learn the techniques of assembling and testing coin cells, which cannot be achieved without numerous help of other researchers in other universities (through frequent calls, email exchanges and two site visits). Thus, we feel that it is beneficial to document, by both text and video, a protocol of assembling and testing a coin cell, which will help other new researchers in this field. This effort represents the "Broader Impact" activities of our NSF project, and it will also help to educate and inspire students. In this video article, we document a protocol to assemble a CR2032 coin cell with a LiCoO2 working electrode, a Li counter electrode, and (the mostly commonly used) polyvinylidene fluoride (PVDF) binder. To ensure new learners to readily repeat the protocol, we keep the protocol as specific and explicit as we can. However, it is important to note that in specific research and development work, many parameters adopted here can be varied. First, one can make coin cells of different sizes and test the working electrode against a counter electrode other than Li. Second, the amounts of C black and binder added into the working electrodes are often varied to suit the particular purpose of research; for example, large amounts of C black or even inert powder were added to the working electrode to test the "intrinsic" performance of cathode materials. Third, better binders (other than PVDF) have also developed and used. Finally, other types of electrolytes (instead of LiPF6) can also be used; in fact, certain high-voltage electrode materials will require the uses of special electrolytes.
Sigma-Aldrich
Poly(vinylidene fluoride), average Mw ~275,000 by GPC, average Mn ~107,000, pellets
Sigma-Aldrich
Poly(vinylidene fluoride), average Mw ~530,000, pellets
Sigma-Aldrich
Poly(vinylidene fluoride), powder
Sigma-Aldrich
Poly(vinylidene fluoride), beads or pellets