Merck
  • Serum proteins of rats exposed to organic solvents examined by horizontal two-dimensional electrophoresis with an immobilized pH gradient in the first dimension.
Serum proteins of rats exposed to organic solvents examined by horizontal two-dimensional electrophoresis with an immobilized pH gradient in the first dimension.
Electrophoresis (1990-08-01)
G Zastrow, S Günther, W Postel, A Görg, H A Diehl, E H Jansen
ABSTRACT
Serum proteins of rats, exposed to methoxyethylacetate and a combination of methoxyethylacetate and isobutylacetate, were analyzed by horizontal high resolution two-dimensional gel electrophoresis with immobilized pH gradients in the first dimension. A total of 410 polypeptides were detected with either increasing or decreasing spot intensities after rat exposure to the organic solvents.
Product Description
Supelco
Isobutyl acetate, analytical standard
Sigma-Aldrich
Isobutyl acetate, natural, ≥97%, FCC, FG
Sigma-Aldrich
Isobutyl acetate, FCC, FG
Supelco
2-Methoxyethyl acetate, suitable for HPLC, ≥99%
Sigma-Aldrich
Isobutyl acetate, 99%