Merck
  • Development of phase-separated scintillators with light-guiding properties.
Development of phase-separated scintillators with light-guiding properties.
Advanced materials (Deerfield Beach, Fla.) (2012-08-14)
Nobuhiro Yasui, Yoshihiro Ohashi, Tamaki Kobayashi, Tohru Den
ABSTRACT
Alkali halide systems that function as phase-separated scintillators (PSSs) with light-guiding properties are sucessfully created. Furthermore, it is the matrix phases of the PSSs which display the light-guiding properties. CsI-NaCl:Tl is a practical material pair because of its high pixel light output and good spatial resolution.
Sigma-Aldrich
Cesium iodide, 99.9% trace metals basis
Sigma-Aldrich
Cesium iodide, 99.999% trace metals basis
Supelco
Cesium iodide, analytical standard, suitable for mass spectrometry (MS)
Sigma-Aldrich
Cesium iodide, AnhydroBeads, −10 mesh, 99.999% trace metals basis