Merck
  • Combination of sulindac and dichloroacetate kills cancer cells via oxidative damage.
Combination of sulindac and dichloroacetate kills cancer cells via oxidative damage.
PloS one (2012-08-07)
Kasirajan Ayyanathan, Shailaja Kesaraju, Ken Dawson-Scully, Herbert Weissbach
ABSTRACT
Sulindac is an FDA-approved non-steroidal anti-inflammatory drug with documented anticancer activities. Our recent studies showed that sulindac selectively enhanced the killing of cancer cells exposed to oxidizing agents via production of reactive oxygen species (ROS) resulting in mitochondrial dysfunction. This effect of sulindac and oxidative stress on cancer cells could be related to the defect in respiration in cancer cells, first described by Warburg 50 years ago, known as the Warburg effect. We postulated that sulindac might enhance the selective killing of cancer cells when combined with any compound that alters mitochondrial respiration. To test this hypothesis we have used dichloroacetate (DCA), which is known to shift pyruvate metabolism away from lactic acid formation to respiration. One might expect that DCA, since it stimulates aerobic metabolism, could stress mitochondrial respiration in cancer cells, which would result in enhanced killing in the presence of sulindac. In this study, we have shown that the combination of sulindac and DCA enhances the selective killing of A549 and SCC25 cancer cells under the conditions used. As predicted, the mechanism of killing involves ROS production, mitochondrial dysfunction, JNK signaling and death by apoptosis. Our results suggest that the sulindac-DCA drug combination may provide an effective cancer therapy.
Product Description
Supelco
Dichloroacetic acid, PESTANAL®, analytical standard
Sigma-Aldrich
Dichloroacetic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Sulindac, ≥98.0%
Sigma-Aldrich
DCA Deblock (0.36M dichloroacetic acid in toluene)